Skip to Content
Merck
All Photos(1)

Key Documents

860817P

Avanti

17:0(2R-OH) Ceramide

Avanti Research - A Croda Brand 860817P, powder

Synonym(s):

N-(2′-(R)-hydroxyheptadecanoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H69NO4
CAS Number:
Molecular Weight:
567.93
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860817P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860817P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCCCCCCCCCCC)=O)CO

InChI

1S/C35H69NO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(38)32(31-37)36-35(40)34(39)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h27,29,32-34,37-39H,3-26,28,30-31H2,1-2H3,(H,36,40)/b29-27+/t32-,33+,34+/m0/s1

InChI key

MBKBHFOACJJHAN-NBVWUYLUSA-N

General description

17:0(2R-OH) Ceramide is a 2R hydroxylated heptadecanoic acid-containing sphingolipid. It is present mainly in epidermis.

Biochem/physiol Actions

Hydroxy fatty acids (hFA)-sphingolipids play a vital role in cell signaling. They also help in cell differentiation and apoptosis. hFA-ceramides are involved in the formation of the epidermal barrier. Fatty acid 2-hydroxylases (FA2H) synthesize hFA-ceramides. Mutations of FA2H have been linked to neurological disorders like spastic paraparesis in humans and leukodystrophy. hFA-ceramides in the cell membrane play a crucial role in imparting the PM02734 drug with anti-tumor activity.

Packaging

5 mL Amber Glass Screw Cap Vial (860817P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2?-Hydroxy ceramide in membrane homeostasis and cell signaling
Kota V and Hama H
Advances in Biological Regulation, 54, 223-230 (2014)
Levels of SCS7/FA2H-mediated fatty acid 2-hydroxylation determine the sensitivity of cells to antitumor PM02734
Herrero AB, et al.
Cancer research, 68(23), 9779-9787 (2008)
Venkatesh Kota et al.
Advances in biological regulation, 54, 223-230 (2013-10-22)
Ceramide is a precursor of complex sphingolipids and also plays important roles in cell signaling. With the advances in lipid analytical technologies, the structural diversity of ceramide species have become evident, and the complexity of cellular metabolism and function associated
Fatty acid 2-Hydroxylation in mammalian sphingolipid biology
Hama H, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1801(4), 405-414 (2010)
Nathan L Alderson et al.
Journal of lipid research, 50(6), 1203-1208 (2009-01-28)
Sphingolipids are ubiquitous components of eukaryotic cells that regulate various cellular functions. In many cell types, a fraction of sphingolipids contain 2-hydroxy fatty acids, produced by fatty acid 2-hydroxylase (FA2H), as the N-acyl chain of ceramide [hydroxyl fatty acid (hFA)-sphingolipids].

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service