Skip to Content
Merck
All Photos(1)

Key Documents

860480P

Avanti

C10 bisphosphonate

Avanti Research - A Croda Brand 860480P, powder

Synonym(s):

1-aminodecane-1,1-bisphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H25NO6P2
CAS Number:
Molecular Weight:
317.26
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 500 μg (860480P-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand 860480P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[NH3+]C(P([O-])(O)=O)(P(O)(O)=O)CCCCCCCCC

InChI

1S/C10H25NO6P2/c1-2-3-4-5-6-7-8-9-10(11,18(12,13)14)19(15,16)17/h2-9,11H2,1H3,(H2,12,13,14)(H2,15,16,17)

InChI key

TWYYYYAFACTHMV-UHFFFAOYSA-N

Application

C10 bisphosphonate is suitable for use as an inhibitor for acid sphingomyelinase.

Biochem/physiol Actions

C10 bisphosphonate, with prolonged carbon chain, is an inhibitor for acid sphingomyelinase and may be a treatment option for inflammatory lung diseases, cystic fibrosis and atherosclerosis. It interacts with the divalent metal ions and forms bidentate complexes. Bisphosphonates are inhibitors of end reaction in pathways. It is an inhibitor of cytosolic farnesyl diphosphate (FDP) synthase (geranyl diphosphate (GDP) and geranylgeranyl diphosphate (GGDP) synthase and isoprene pathway. Bisphosphonates have antimicrobial functionality and inhibits the growth of Entamoeba histolytica and Plasmodium falciparum.

Packaging

5 mL Amber Glass Screw Cap Vial (860480P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volker Teichgräber et al.
Nature medicine, 14(4), 382-391 (2008-04-01)
Microbial lung infections are the major cause of morbidity and mortality in the hereditary metabolic disorder cystic fibrosis, yet the molecular mechanisms leading from the mutation of cystic fibrosis transmembrane conductance regulator (CFTR) to lung infection are still unclear. Here
Bisphosphonate inhibitors reveal a large elasticity of plastidic isoprenoid synthesis pathway in isoprene-emitting hybrid aspen
Rasulov B, et al.
Plant Physiology, 168(2), 532-548 (2015)
Effects of bisphosphonates on the growth of Entamoeba histolytica and Plasmodium species in vitro and in vivo
Ghosh S, et al.
Journal of Medicinal Chemistry, 47(1), 175-187 (2004)
Acid sphingomyelinase promotes mitochondrial dysfunction due to glutamate-induced regulated necrosis
Novgorodov SA, et al.
Journal of Lipid Research, 59(2), 312-329 (2018)
Potent and selective inhibition of acid sphingomyelinase by bisphosphonates
Roth AG, et al.
Angewandte Chemie (International Edition in English), 48(41), 7560-7563 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service