Skip to Content
Merck
All Photos(2)

Key Documents

860355C

Avanti

16:0 PC-d62

Avanti Research - A Croda Brand 860355C

Synonym(s):

1,2-dipalmitoyl-d62-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H18NO8PD62
CAS Number:
Molecular Weight:
796.42
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.12

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860355C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860355C

concentration

10 mg/mL (860355C-10mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)COC(C([2H])([2H

InChI

1S/C40H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-39(42)46-36-38(37-48-50(44,45)47-35-34-41(3,4)5)49-40(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3/t38-/m1/s1/i1D3,2D3,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2,30D2,31D2,32D2,33D2

InChI key

KILNVBDSWZSGLL-PWXLRKPBSA-N

General description

16:0 PC-d62 is a deuterated phosphocholine (PC) in which 62 protons of dipalmitoyl are replaced by deuterium.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0 PC-d62 may be used for the preparation of asymmetric large unilamellar vesicles (aLUVs) for use as cell membrane models.

Packaging

5 mL Clear Glass Sealed Ampule (860355C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of asymmetric phospholipid vesicles for use as cell membrane models
Doktorova M, et al.
Nature Protocols, 13(9), 2086-2101 (2018)
Kalliopi-Kelli A Vandera et al.
Molecular pharmaceutics, 17(7), 2354-2369 (2020-05-01)
Gram-negative bacteria possess numerous defenses against antibiotics, due to the intrinsic permeability barrier of their outer membrane (OM), explaining the recalcitrance of some common and life-threatening infections. We report the formulation of a new drug, PPA148, which shows promising activity
Benjamin Martial et al.
The journal of physical chemistry. B, 124(17), 3469-3481 (2020-04-02)
α-Synuclein (AS), the protein responsible for Parkinson's disease, contains a 12-residue-long sequence, AS71-82, that is thought to play a crucial role in the α-synuclein aggregation process. Neuronal membranes are direct interacting partners of α-synuclein and play a role in fibrillogenesis
Drew Marquardt et al.
Langmuir : the ACS journal of surfaces and colloids, 33(15), 3731-3741 (2017-01-21)
We measured the transbilayer diffusion of 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) in large unilamellar vesicles, in both the gel (Lβ') and fluid (Lα) phases. The choline resonance of headgroup-protiated DPPC exchanged into the outer leaflet of headgroup-deuterated DPPC-d13 vesicles was monitored using 1H
Mitchell DiPasquale et al.
Biochimica et biophysica acta. Biomembranes, 1862(8), 183189-183189 (2020-01-19)
The antioxidant vitamin E is a commonly used vitamin supplement. Although the multi-billion dollar vitamin and nutritional supplement industry encourages the use of vitamin E, there is very little evidence supporting its actual health benefits. Moreover, vitamin E is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service