Skip to Content
Merck
All Photos(2)

Key Documents

855776P

Avanti

19:0 Lyso PC

Avanti Research - A Croda Brand 855776P, powder

Synonym(s):

1-nonadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H56NO7P
CAS Number:
Molecular Weight:
537.71
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 200 mg (855776P-200mg)
pkg of 1 × 25 mg (855776P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855776P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCCCCC)=O

InChI

1S/C27H56NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-27(30)33-24-26(29)25-35-36(31,32)34-23-22-28(2,3)4/h26,29H,5-25H2,1-4H3/t26-/m1/s1

InChI key

WYIMORDZGCUBAA-AREMUKBSSA-N

Application

19:0 Lyso PC has been used as an internal standard to measure endogenous phosphatidylcholine (PC) in different types of uterine cells using quantitative nanospray desorption electrospray ionization mass spectrometry imaging (nano-DESI MSI).

Packaging

5 mL Amber Glass Screw Cap Vial (855776P-200mg)
5 mL Amber Glass Screw Cap Vial (855776P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruichuan Yin et al.
Nature protocols, 14(12), 3445-3470 (2019-11-15)
Mass spectrometry imaging (MSI) enables label-free spatial mapping of hundreds of biomolecules in tissue sections. This capability provides valuable information on tissue heterogeneity that is difficult to obtain using population-averaged assays. Despite substantial developments in both instrumentation and methodology, MSI
High spatial resolution imaging of biological tissues using nanospray desorption electrospray ionization mass spectrometry
Yin R, et al.
Nature Protocols, 14(12), 3445-3470 (2019)
Sidra Iqbal et al.
Plant physiology and biochemistry : PPB, 147, 101-112 (2019-12-20)
Plant phospholipase A (PLA) and C (PLC) families are least explored in terms of structure, diversity and their roles in membrane lipid remodeling under stress conditions. In this study, we performed gene family analysis, determined gene expression in different tissues
Marco Custódio et al.
Scientific reports, 10(1), 6583-6583 (2020-04-22)
Halophytes are increasingly regarded as suitable extractive species and co-products for coastal Integrated Multi-Trophic Aquaculture (IMTA) and studying their lipidome is a valid means towards their economic valorization. Halimione portulacoides (L.) Aellen edible leaves are rich in functional lipids with
Jinfeng Huang et al.
Dose-response : a publication of International Hormesis Society, 18(2), 1559325820914209-1559325820914209 (2020-05-05)
Radiation biodosimeters are required urgently for fast and accurate evaluation of absorbed dose for irradiated individuals. Lipidomics has appeared as a credible technique for identification and quantification of lipid for researching biomarker of diseases. We performed a lipidomic profile on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service