Skip to Content
Merck
All Photos(2)

Key Documents

855675C

Avanti

16:0 Lyso PC

1-palmitoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-hexadecanoyl-sn-glycero-3-phosphocholine; PC(16:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H50NO7P
CAS Number:
Molecular Weight:
495.63
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855675C-25mg)
pkg of 2 × 4 mL (855675C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855675C

concentration

10 mg/mL (855675C-25mg)
25 mg/mL (855675C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C24H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(27)30-21-23(26)22-32-33(28,29)31-20-19-25(2,3)4/h23,26H,5-22H2,1-4H3/t23-/m1/s1

InChI key

ASWBNKHCZGQVJV-HSZRJFAPSA-N

Application

16:0 Lyso PC has been used to reveal the central role of the adipocyte inflammasome in homocysteine-induced insulin resistance. It has also been used for lipid extraction and mass spectrometry analysis.

Packaging

5 mL Clear Glass Sealed Ampule (855675C-200mg)
5 mL Clear Glass Sealed Ampule (855675C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Song-Yang Zhang et al.
EBioMedicine, 31, 202-216 (2018-05-08)
The adipose Nod-like receptor protein 3 (NLRP3) inflammasome senses danger-associated molecular patterns (DAMPs) and initiates insulin resistance, but the mechanisms of adipose inflammasome activation remains elusive. In this study, Homocysteine (Hcy) is revealed to be a DAMP that activates adipocyte
Takafumi Enomoto et al.
Langmuir : the ACS journal of surfaces and colloids, 34(3), 750-755 (2017-10-06)
A major goal of synthetic biology is the development of rational methodologies to construct self-assembling non-natural membranes, which could enable the efficient fabrication of artificial cellular systems from purely synthetic components. However, spatiotemporal control of artificial membrane formation remains both
Xinyuan Li et al.
Redox biology, 28, 101373-101373 (2019-11-16)
It has been shown that anti-inflammatory cytokines interleukin-35 (IL-35) and IL-10 could inhibit acute endothelial cell (EC) activation, however, it remains unknown if and by what pathways IL-35 and IL-10 could block atherogenic lipid lysophosphatidylcholine (LPC)-induced sustained EC activation; and
Li V Yang et al.
Blood, 105(3), 1127-1134 (2004-09-24)
G2A is a G-protein-coupled receptor (GPCR) involved in immune regulation. Previous studies have shown that lysophosphatidylcholine (LPC), a bioactive lipid associated with atherosclerosis and autoimmunity, acts through G2A to induce diverse biologic effects. Production of LPC during cell apoptosis serves
Oxidized phospholipids inhibit the formation of cholesterol-dependent plasma membrane nanoplatforms
Brameshuber M, et al.
Biophysical Journal, 110(1), 205-213 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service