Skip to Content
Merck
All Photos(2)

Key Documents

850373C

Avanti

24:0 PC

1,2-dilignoceroyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-ditetracosanoyl-sn-glycero-3-phosphocholine; PC(24:0/24:0) ; 110929

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H112NO8P
CAS Number:
Molecular Weight:
958.46
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850373C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850373C

concentration

10 mg/mL (850373C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

24:0 PC (1,2-dilignoceroyl-sn-glycero-3-phosphocholine) is a long-chain diacyl phospholipid, which has C24 saturated hydrocarbon chains. This lipid has a phosphatidylcholine (PC) headgroup.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

24:0 PC (1,2-dilignoceroyl-sn-glycero-3-phosphocholine) may be used:
  • in short-chain micelles, longer-chain oriented multilayers and in Langmuir monolayers for structural studies
  • as an acyl chain lipid in microbubble condensed droplets (MCDs) and characterization studies
  • in phosphatidylcholine bilayer preparation for Raman spectroscopy studies

Biochem/physiol Actions

Phosphatidylcholine (PC) can prevent the penetrance of bacteria by acting as a surfactant. This phospholipid is capable of forming a strong bilayer.

Packaging

5 mL Clear Glass Sealed Ampule (850373C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Dmitriev et al.
The journal of physical chemistry. B, 119(51), 15613-15622 (2015-11-27)
Raman scattering of five phosphatidylcholines [1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC), 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine (DPPC), 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC), 1,2-dilinoleoyl-sn-glycero-3-phosphocholine (DLPC), and 1,2-dilignoceroyl-sn-glycero-3-phosphocholine (PC24)] was studied in a wide temperature range. These phospholipid bilayers differ in the temperature of the gel-fluid transition Tm (from -57 to +80 °C)
Inflammatory Bowel Disease
Parian AM, et al.
Integrative Medicine, 501-516 (2018)
S Zheng et al.
Biophysical journal, 80(4), 1837-1850 (2001-03-22)
Vpu is an 81 amino acid integral membrane protein encoded by the HIV-1 genome with a N-terminal hydrophobic domain and a C-terminal hydrophilic domain. It enhances the release of virus from the infected cell and triggers degradation of the virus
Lipid-Protein Interactions in Membranes
The Membranes of Cells, 291-334 (2016)
Paul A Mountford et al.
Langmuir : the ACS journal of surfaces and colloids, 31(16), 4627-4634 (2015-04-09)
This study explored the thermal conditions necessary for the vaporization of superheated perfluorocarbon nanodrops. Droplets C3F8 and C4F10 coated with a homologous series of saturated diacylphosphatidylcholines were formed by condensation of 4 μm diameter microbubbles. These drops were stable at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service