Skip to Content
Merck
All Photos(1)

Key Documents

710335P

Avanti

18:1 Cardiolipin

Avanti Research - A Croda Brand

Synonym(s):

tetraoleoyl cardiolipin; 1,1′,2,2′-tetra-(9Z-octadecenoyl) cardiolipin (sodium salt); CL(1′-[18:1(9Z)/18:1(9Z)],3′-[18:1(9Z)/18:1(9Z)])

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C81H148O17P2Na2
CAS Number:
Molecular Weight:
1501.96
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1′,3′-bis[1,2-dioleoyl-sn-glycero-3-phospho]-glycerol (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (710335P-200mg)
pkg of 5 × 100 mg (710335P-500mg)
pkg of 1 × 25 mg (710335P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

Cardiolipin has two phosphatidyl residues linked by glycerol. It is present in the bacterial membranes.[1] 18:1 cardiolipin or tetraoleoyl cardiolipin (TOCL) is a monounsaturated[2] mitochondrial phospholipid.[3] The bilayer thickness of TOCL is higher than the heart cardiolipin.[2] TOCL bilayers have a larger hydrocarbon chain thickness compared to dioleoyl phosphatidylcholine (DOPC) bilayer.[4]
Cardiolipin is a unique phospholipid located in the inner mitochondrial membrane,[5] which makes up to 20% of total lipids.[6] Cardiolipin is a non-bilayer anionic phospholipid, which has four acyl chains and a small headgroup.[7]
Cardiolipin is an anionic phospholipid.[7]

Application

18:1 Cardiolipin has been used as a lipid substrate for luminol-enhanced chemiluminescence.[8][9]
18:1 Cardiolipin has been used:
  • to prepare pure (100%) cardiolipin vesicles [10]
  • to measure the intensity of cytochrome c peroxidase activity by luminescence assay[11]
  • to measure the fluorescence of cytochrome c Trp59[11] immobilized in biosensor chip
  • to study the association of cytochrome c with anionic phospholipids[11]

18:1 Cardiolipin has been used:
  • in the preparation of liposomes for reconstitution of mechanosensitive channels from bacteria[1]
  • in the immobilization on lipophilic biosensor L1 chip surface[8]
  • as lipid substrate in peroxidase oxidation assay[9]

Biochem/physiol Actions

18:1 Cardiolipin or tetraoleoyl cardiolipin (TOCL) regulates oxidative phosphorylation and respiration in mitochondria.[3] Metabolic pathway defects in cardiolipin is associated with Barth syndrome patients.[2] TOCL undergoes oxidation in the presence of cytochrome C.[9]
Cardiolipin and cytochrome c interaction determines the function of proteins involved in oxidative phosphorylation and apoptosis.[12] Cardiolipin involved in proton uptake pathway maintains the structure of enzymes participating.[7] Cardiolipin controls the mitochondrial function and membrane potential. It offers structural support for the inner mitochondrial membrane and proteins in it. Altered cardiolipin concentration and composition have been implicated in pathological conditions including ischemia, hypothyroidism, aging, heart failure and cardioskeletal myopathy.
Cardiolipin is involved in the dynamic organization of membranes and in cell division.[13] It regulates the proton uptake pathway.[7] Cardiolipin plays a critical role in mitochondrial biogenesis, fusion and fission, respiration and protein import. It controls cell wall biogenesis, vacuole homeostasis, aging, cell cycle and apoptosis.[14]

Packaging

5 mL Clear Glass Sealed Ampule (710335P-200mg)
5 mL Clear Glass Sealed Ampule (710335P-25mg)
5 mL Clear Glass Sealed Ampule (710335P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

755.6 - 820.4 °F

Flash Point(C)

402 - 438 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of cytochrome c affinity to anionic phospholipids by means of surface plasmon resonance
Stepanov G, et al.
Febs Letters, 97?100-97?100 (2009)
The role of cardiolipin in cardiovascular health
Shen Z, et al.
BioMed Research International, 2015 (2015)
Patch clamp characterization of the effect of cardiolipin on MscS of E. coli
Ridone P, et al.
European Biophysics Journal, 44(7), 567-576 (2015)
Lipidomics: Technologies and applications (2012)
Lipoperoxide radical production during oxidation of cardiolipin in the complex with cytochrome c
Vladimirov YA, et al.
Biochemistry (Moscow) Supplement Series A: Membrane and Cell Biology, 3(4), 467-477 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service