Skip to Content
Merck
All Photos(1)

Key Documents

791268P

Avanti

DTPA-BSA (Gd)

DTPA-bis(stearylamide) (gadolinium salt), powder

Synonym(s):

(diethylenetriaminepentaacetic acid)-bis(stearylamide) (gadolinium salt); Gd-DTPA-BSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H94GdN5O8
CAS Number:
Molecular Weight:
1050.56
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (791268P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791268P

shipped in

dry ice

storage temp.

−20°C

General description

DTPA-BSA (Gd) (diethylenetriaminepentaacetic acid)-bis(stearylamide) (gadolinium salt)) is an amphiphilic molecule with paramagnetic property.

Application

DTPA-BSA (Gd) (diethylenetriaminepentaacetic acid)-bis(stearylamide) (gadolinium salt)) has been used:
  • in the preparation of nontargeting (NT) gadolinium peptide amphiphile micelles (PAMs) for the magnetic resonance imaging (MRI) of atherosclerotic lesions
  • as a MRI agent in the preparation of lipid-like nanoparticles (LLNs) for small interfering RNA (siRNA) encapsulation
  • as a MRI agent in the preparation of paramagnetic saposin C and dioleylphosphatidylserine (SapC-DOPS) vesicles for imaging glioblastoma multiforme (GBM) tumors

Biochem/physiol Actions

Gd-DTPA-BSA is useful as magnetic resonance imaging (MRI) agent and the gadolinium ion enables the generation of bright images by lowering the longitudinal relaxation time.

Packaging

5 mL Amber Glass Screw Cap Vial (791268P-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Jasanada et al.
Bioconjugate chemistry, 7(1), 72-81 (1996-01-01)
In order to use the LDL receptor pathway to target radionuclides to cancer sites for imaging and diagnostic purposes, a labeling procedure of LDL with 111In using the DTPA-bis(stearylamide) (L) has been developed. This bifunctional ligand is intended to be
Dual-functional lipid-like nanoparticles for delivery of mRNA and MRI contrast agents
Luo X, et al.
Nanoscale, 9(4), 1575-1579 (2017)
Sang Pil Yoo et al.
ACS omega, 1(5), 996-1003 (2016-12-06)
The leading causes of morbidity and mortality globally are cardiovascular diseases, and nanomedicine can provide many improvements including disease-specific targeting, early detection, and local delivery of diagnostic agents. To this end, we designed fibrin-binding, peptide amphiphile micelles (PAMs), achieved by
X Luo et al.
Nanoscale, 9(4), 1575-1579 (2017-01-10)
Multi-functional nanomaterials possess unique properties, facilitating both therapeutic and diagnostic applications among others. Herein, we developed dual-functional lipid-like nanoparticles for simultaneous delivery of mRNA and magnetic resonance imaging (MRI) contrast agents in order to express functional proteins and provide real-time
Geralda A F van Tilborg et al.
Bioconjugate chemistry, 17(3), 741-749 (2006-05-18)
Apoptosis, or programmed cell death, plays an important role in the etiology of a variety of diseases, including cancer and myocardial infarction. Visualization of apoptosis would allow both early detection of therapy efficiency and evaluation of disease progression. To that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service