Skip to Content
Merck
All Photos(1)

Key Documents

86734

Sigma-Aldrich

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid

≥97.0% (CHN)

Synonym(s):

DOTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N4O8 · xH2O
CAS Number:
Molecular Weight:
404.42 (anhydrous basis)
Beilstein:
1186987
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (CHN)

form

powder

functional group

carboxylic acid

SMILES string

OC(=O)CN1CCN(CCN(CCN(CC1)CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C16H28N4O8/c21-13(22)9-17-1-2-18(10-14(23)24)5-6-20(12-16(27)28)8-7-19(4-3-17)11-15(25)26/h1-12H2,(H,21,22)(H,23,24)(H,25,26)(H,27,28)

InChI key

WDLRUFUQRNWCPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTA) is a macrocyclic complexing agent.
  • It has been used for radiolabeling of carbon nanotube bioconjugates by chelating 64Cu radioisotope.
  • DOTA can be activated with N-hydroxysulfosuccinimidyl for conjugation with monoclonal antibodies in clinical radioimmunotherapy.
  • It can form complexes with gadolinium for application as MRI contrast agents.
  • Metal complexes of DOTA-peptide conjugates can be used as therapeutic radiopharmaceuticals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved method for conjugating monoclonal antibodies with N-hydroxysulfosuccinimidyl DOTA.
Lewis MR, et al.
Bioconjugate Chemistry, 12(2), 320-324 (2001)
The synthesis and chelation chemistry of DOTA- peptide conjugates.
De Leon-Rodriguez LM and Kovacs Z
Bioconjugate Chemistry, 19(2), 391-402 (2007)
Sara S Rinne et al.
International journal of molecular sciences, 21(6) (2020-03-19)
HER3-binding affibody molecules are a promising format for visualization of HER3 expression. Cobalt-55, a positron-emitting isotope, with a half-life of 17.5 h, allows for next-day imaging. We investigated the influence of the charge of the radiocobalt-chelator complex on the biodistribution
Luca Garbuio et al.
The journal of physical chemistry. B, 117(11), 3145-3153 (2013-02-28)
We present the first example of chemoselective site-specific spin labeling of a monomeric protein with two spectroscopically orthogonal spin labels: a gadolinium(III) chelate complex and a nitroxide radical. A detailed analysis of the performance of two commercially available Gd(III) ligands
Jack J Miller et al.
NMR in biomedicine, 31(6), e3912-e3912 (2018-04-11)
The aim of this work was to investigate the use of 13 C-labelled acetoacetate and β-hydroxybutyrate as novel hyperpolarized substrates in the study of cardiac metabolism. [1-13 C]Acetoacetate was synthesized by catalysed hydrolysis, and both it and [1-13 C]β-hydroxybutyrate were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service