Skip to Content
Merck
All Photos(1)

Key Documents

750332C

Avanti

14:0 Cardiolipin (sodium salt)

1′,3′-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-glycerol (sodium salt), chloroform

Synonym(s):

1,1′,2,2′-tetratetradecanoyl cardiolipin (sodium salt); CL(1′-[14:0/14:0],3′-[14:0/14:0])

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H124Na2O17P2
CAS Number:
Molecular Weight:
1285.60
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

solution

packaging

pkg of 1 × 2.5 mL (750332C-25mg)
pkg of 2 × 4 mL (750332C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 750332C

concentration

10 mg/mL (750332C-25mg)
25 mg/mL (750332C-200mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

Cardiolipin, the mitochondrial-specific phospholipid, helps maintain mitochondrial function, membrane potential and structural support for the inner mitochondrial membrane and the proteins in it. Concentration and composition changes of cardiolipin have been implicated in pathological conditions including ischemia, hypothyroidism, aging, heart failure and cardioskeletal myopathy (Barth syndrome). Cardiolipin has also been shown as a signaling molecule in apoptosis.

Application

14:0 Cardiolipin (sodium salt) has been used for the mass spectrometry (MS) analysis to determine the structure of lipid A.

Packaging

5 mL Clear Glass Sealed Ampule (750332C-200mg)
5 mL Clear Glass Sealed Ampule (750332C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid lipid a structure determination via surface acoustic wave nebulization and hierarchical tandem mass spectrometry algorithm
Yoon SH, et al.
Rapid Communications in Mass Spectrometry, 30(23), 2555-2560 (2016)
Role of cardiolipin alterations in mitochondrial dysfunction and disease
Chicco AJ and Sparagna GC
American Journal of Physiology. Cell Physiology, 292(1), C33-C44 (2007)
Vladimir B Ritov et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 831(1-2), 63-71 (2005-12-13)
Cardiolipin is a phospholipid that is specific to the inner mitochondrial membrane and essential for numerous mitochondrial functions. Accordingly, a quantitative assay for cardiolipin can be a valuable aspect of assessing mitochondrial content and functional capacity. The current study was
Yuchen Sun et al.
Metabolites, 9(11) (2019-11-07)
Extracellular vesicles (EVs) consist of lipid bilayers, occur in various biofluids, and are invaluable in biomarker screening. Liquid chromatography coupled with high-resolution mass spectrometry (LC-MS) was recently used to study comprehensive EV lipid profiles in vitro. The aim of this
Na Li et al.
Analytical chemistry, 91(24), 15873-15882 (2019-11-14)
The matrix plays a prominent role in expanding the ability of matrix assisted laser desorption/ionization mass spectrometry (MALDI MS). However, on account of the unclarity of necessary properties of the matrix in MALDI MS, development of a new matrix is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service