Skip to Content
Merck
All Photos(1)

Key Documents

341602G

Avanti

L-α-Phosphatidylcholine

40% (Soy), Avanti Polar Lipids

Synonym(s):

L-α-Phosphatidylcholine (Soy-40%)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Product Name

Soy PC (40%), Avanti Research - A Croda Brand

form

granular

packaging

pkg of 1 × 50 g (341602G-50g)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Soy phosphatidylcholine (PC) or lecithin is isolated from Glycine Max (soybean). It is a phospholipid mixture with phosphatidylcholine as a major component, followed by phosphatidylethanolamine and phosphatidylinositol.

Application

Soy PC (40%) has been used in the preparation of cytochrome c-encapsulated liposomes for proteoliposomes internalization studies.

Biochem/physiol Actions

Soy phosphatidylcholine (PC) is rich in polyunsaturated fatty acids (PUFAs) and is used to mimic biological membranes. It has wide applications as an emulsifier in the food, pharmaceuticals and cosmetics industries. Soy PC based liposomes are useful as a carrier for anticancer drug, paclitaxel.

Packaging

125 mL Amber Wide Mouth Glass Bottle with Screw Cap (341602G-50g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of paclitaxel-loaded liposomal nanocarrier stabilized by triglyceride incorporation
Hong SS, et al.
International journal of nanomedicine, 11, 4465-4465 (2016)
Sergey Deyev et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 130, 296-305 (2018-07-01)
Since their discovery, liposomes have been widely employed in biomedical research. These nano-size spherical vesicles consisting one or few phospholipid bilayers surrounding an aqueous core are capable of carrying a wide variety of bioactive compounds, including drugs, peptides, nucleic acids
Soybean phosphatidylcholine liposomes as model membranes to study lipid peroxidation photoinduced by pterin
Thomas AH, et al.
Biochimica et Biophysica Acta - Biomembranes, 1858(1), 139-145 (2016)
Behavioral effect of phosphatidylcholine isolated from soy lecithin in streptozotocin induced experimental alzheimers model
Gulkari VD, et al.
Journal of Pharmacognosy and Phytochemistry, 6(3), 702-709 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service