Z742526
Block for digital dry bath and MyBlock™
Holds 1 x 96-well PCR plate
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NB.22
Recommended Products
feature
Holds 1 x 96-well PCR plate
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BSWPCR1
technique(s)
PCR: suitable
Related Categories
General description
For use with Benchmark Digital Dry Bath
- Material: high grade aluminum
- Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Legal Information
MyBlock is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)