Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Block for digital dry bath and MyBlock
™
Products
Manufacturer
Package Size
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Z742523
Block for digital dry bath and MyBlock
™
,
Holds 5 x 50 mL tubes
Expand
View Pricing
Z742517
Block for digital dry bath and MyBlock
™
,
Holds 24 x 1.5 mL or 2.0 x tubes
Expand
View Pricing
Z742520
Block for digital dry bath and MyBlock
™
,
Holds 12 x 15 mL tubes
Expand
View Pricing
Z742516
Block for digital dry bath and MyBlock
™
,
Holds 24 x 1.5 mL tubes
Expand
View Pricing
Z742524
Block for digital dry bath and MyBlock
™
,
Holds 12 x 15-16 mm tubes
Expand
View Pricing
Z742522
Block for digital dry bath and MyBlock
™
,
Holds 20 x 12-13 mm tubes
Expand
View Pricing
Z742514
Block for digital dry bath and MyBlock
™
,
solid block for slides or custom machining
Expand
View Pricing
Z742521
Block for digital dry bath and MyBlock
™
,
for 20 x 10 mm test tubes or 20 x 2.0 mL centrifuge tubes
Expand
View Pricing
Z742527
Block for digital dry bath and MyBlock
™
,
Holds 1 x 96-well PCR plate
Expand
View Pricing
Z742525
Block for digital dry bath and MyBlock
™
,
for 24 x 1.5 mL tubes, or 32 x 0.2 mL and 14 x 0.5 mL tubes
Expand
View Pricing
Z742526
Block for digital dry bath and MyBlock
™
,
Holds 1 x 96-well PCR plate
Expand
View Pricing
Z742519
Block for digital dry bath and MyBlock
™
,
Holds 12 x 5.0 mL tubes
Expand
View Pricing
Z742515
Block for digital dry bath and MyBlock
™
,
Holds 48 x 0.2 mL
Expand
View Pricing
Z742518
Block for digital dry bath and MyBlock
™
,
Holds 24 x 0.5 mL tubes
Expand
View Pricing
Z742528
Block for digital dry bath and MyBlock
™
,
for 2 or 4 block models, Holds 12 x 5 mL centrifuge tubes (17mm diameter) OR microplate, skirted or non-skirted
Expand
View Pricing