Z645265
IKA® sieves for MF 10 basic microfine grinder drive
MF 0.25, 0.25mm, 1/cs
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
Recommended Products
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 2938900
size
0.25 mm
Related Categories
General description
Note: Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Alexey Veremeev et al.
Materials (Basel, Switzerland), 13(15) (2020-08-06)
Here we evaluated the efficacy of bone repair using various native bovine biomaterials (refined hydroxyapatite (HA), demineralised bone matrix (DBM), and purified bone collagen (COLL)) as compared with commercially available bone mineral and bone autografts. We employed a conventional critical-sized