Z533017
Aldrich® three-neck flask with bottom outlet
capacity 500 mL, joint: ST/NS 24/40
Synonym(s):
bottom flask, flask round, flask round bottom
About This Item
Recommended Products
material
borosilicate glass
round bottom
three-neck
three-neck flask
Joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
outlet fitting size
4 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service