Skip to Content
Merck
All Photos(2)

Key Documents

Z273228

Sigma-Aldrich

Alconox® detergent

0.5 oz packs

Synonym(s):

anionic detergent, concentrated powdered detergent, general laboratory cleaner, lab detergent, low foaming detergent, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47131800
NACRES:
NB.35

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1613
suitable for EPA 601
suitable for EPA 8327
suitable for SM 5210

packaging

pkg of 12 x 50 × 0.5 oz in 12PAK
pkg of 50 × 0.5 oz in 1PAK

manufacturer/tradename

Alconox, Inc. 1112 (12 packs of 50ea)
Alconox, Inc. 1112-1 (1 pack of 50ea)

application(s)

PFAS testing

Looking for similar products? Visit Product Comparison Guide

General description

Alconox® detergent is applicable for cleaninglabware, healthcare instruments, tissue culture ware, catheters, tubing, pipes,personal protective equipment, vacuum equipment, and sampling apparatus. It isalso suitable to clean radioactive contaminated articles, electroniccomponents, pharmaceutical apparatus, optical parts, metal castings, forgingsand stampings, industrial parts, tanks, reactors, and cosmetics manufacturingequipment. Alconox detergent is suitable to eliminate grime, soil, slime,blood, grease, oils, deposits, tissues, particulates, chemicals, grit,radioactive contaminations, mold release agents, silicon oils, andradioisotopes.
Powdered detergent for general lab cleaning.

Application

ALCONOX® detergent has been used:

  • to clean tissue grinder
  • for pre-treating glass coverslips
  • microfluidic chambers

Features and Benefits

  • Highly concentrated and cost-efficient
  • Biodegradable and readily disposable
  • Penetrating wetting power saves time
  • Provides reliable results and no interfering residues
  • Alternative for corrosive acids and hazardous solvents

Legal Information

ALCONOX is a registered trademark of Alconox, Inc.
ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marco Spinazzi et al.
Nature protocols, 7(6), 1235-1246 (2012-06-02)
The assessment of mitochondrial respiratory chain (RC) enzymatic activities is essential for investigating mitochondrial function in several situations, including mitochondrial disorders, diabetes, cancer, aging and neurodegeneration, as well as for many toxicological assays. Muscle is the most commonly analyzed tissue
Mengyu Liu et al.
Autophagy, 17(4), 903-924 (2020-03-12)
Trimethyltin chloride (TMT) is widely used as a constituent of fungicides and plastic stabilizers in the industrial and agricultural fields, and is generally acknowledged to have potent neurotoxicity, especially in the hippocampus; however, the mechanism of induction of neurotoxicity by
Michelle Richner et al.
Nature protocols, 10(10), 1543-1555 (2015-09-18)
The ability to generate human neurons of specific subtypes of clinical importance offers experimental platforms that may be instrumental for disease modeling. We recently published a study demonstrating the use of neuronal microRNAs (miRNAs) and transcription factors to directly convert

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service