Skip to Content
Merck
All Photos(2)

Key Documents

Z273198

Sigma-Aldrich

Alcojet® detergent

greener alternative

pkg of 1.8 kg

Synonym(s):

biodegradable cleaner, low-foaming powdered cleaner, non-ionic detergent, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

description

non-ionic

packaging

pkg of 1.8 kg

manufacturer/tradename

Alconox, Inc. 1404-1 (1.8 KG)

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

Alcojet is a low-foaming powdered detergent

Used to clean: Dissection instruments, laboratory ware, sampling apparatus, plate glass, pharmaceutical apparatus, cosmetics manufacturing equipment, restaurant equipment, wine glasses, counters, floors, sidewalks and driveways, metal castings, forgings and stampings, industrial parts, tanks and reactors. Authorized by USDA for use in federally inspected meat and poultry plants. Passes inhibitory residue test for water analysis. FDA certified.

Used to remove: Soil, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, inorganic residues, particulates, solvents, chemicals and solvents.

Surfaces cleaned: Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, grout, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Corrosion testing may be advisable.



Cleaning method:
Laboratory warewashers, cart washers, cage washers, spray washers, electrocleaning, ultrasonic cleaners, flow through clean-in-place. Manual cleaning if proper skin and eye protection is used.

Directions: Follow machine manufacturer′s directions and use machine′s powder detergent receptacles. In general, use 1/2 - 1% hot water wash cycle solutions (1 1/4 - 2 1/2 Tbsp. per gal., 3/4 - 1 1/4 oz. per gal. or 5-10 grams per liter. Most machines need 2 1/2 oz. added at a typical 2 gal. wash cycle).


For manual, soak, circulate, or ultrasonic use, make a 1% solution in warm or hot water.


For abrasive scrubbing, make paste.


For difficult soils, raise water temperature and use more detergent.


Wear protective gloves and eyewear for manual cleaning.


RINSE THOROUGHLY preferably with running water. For critical cleaning, do final or all rinsing in distilled, deionized, or purified water. For food contact surfaces, rinse with potable water.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Features and Benefits

  • Concentrated to save you money
  • Biodegradable and readily disposable
  • Non-ionic, low foaming cleaner for high performance from your cleaning machinery without damaging washer parts
  • Free rinsing to give you reliable results and no interfering residues
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval

Legal Information

Alcojet is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service