Z247243
Coors™ porcelain crucible cover
cap size E, I.D. 74 mm
All Photos(1)
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
Recommended Products
cap size
E
packaging
pkg of 10 ea
manufacturer/tradename
Coorstek 66126
I.D.
74 mm
General description
Coors™ porcelain crucible cover is glazed all over except for the rim.
Features and Benefits
- Ensures great resistance to thermal-shock
- Durable
- Rigid
- Sustained refractory qualities
Other Notes
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)