Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Coors
™
porcelain crucible cover
Products
Product Category
crucibles (6)
Manufacturer
ID (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z247235
Coors
™
porcelain crucible cover,
cap size D, I.D. 54 mm
Expand
View Pricing
Z247243
Coors
™
porcelain crucible cover,
cap size E, I.D. 74 mm
Expand
View Pricing
Z247227
Coors
™
porcelain crucible cover,
cap size C, I.D. 49
Expand
View Pricing
Z247251
Coors
™
porcelain crucible cover,
cap size F, I.D. 84 mm
Expand
View Pricing
Z247200
Coors
™
porcelain crucible cover,
cap size A, I.D. 34 mm
Expand
View Pricing
Z247219
Coors
™
porcelain crucible cover,
cap size B, I.D. 39 mm
Expand
View Pricing