Skip to Content
Merck
All Photos(1)

Key Documents

Z176990

Measure-Matic dispenser

measures from 5 to 25 mL, polyethylene/polyallomer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

Pricing and availability is not currently available.

material

polypropylene

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F11654-0000

volume measuring range

5-25 mL

volume

5-25 mL

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4503468SAB2109100HPA055087
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

-

product line

-

product line

Prestige Antibodies® Powered by Atlas Antibodies

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:200-1:500, immunofluorescence: 0.25-2 μg/mL

technique(s)

ELISA: 1:40000, immunohistochemistry: 1:50-1:100, western blot: 1:500-1:1000

technique(s)

western blot: 1 μg/mL

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:1000-1:2500

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

PE/polyallomer. Measures any amount from 5 to 25 mL; fills from 16 oz PE storage bottle. Pressure on bottle forces liquid above holes in tube. Removal of pressure allows return of excess liquid to storage bottle. Supplied with hollow stopper as dust cover.

Legal Information

Measure-Matic is a trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service