Skip to Content
Merck
All Photos(1)

Key Documents

W507709

Sigma-Aldrich

L-Fenchone

≥98%

Synonym(s):

(1R)-(−)-Fenchone, (−)-1,3,3-Trimethyl-2-norbornanone, (−)-Fenchone, (1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
4519
Beilstein:
2042710
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009

Assay

≥98%

optical activity

[α]20/D −51°, neat

refractive index

n20/D 1.461 (lit.)

bp

192-194 °C (lit.)

mp

5-6 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

camphoraceous; earthy; woody

SMILES string

CC1(C)C2CCC(C)(C2)C1=O

InChI

1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3/t7-,10+/m0/s1

InChI key

LHXDLQBQYFFVNW-OIBJUYFYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Fenchone is a monoterpene that occurs in the essential oils of plants such as Lavandula dentate. It shows potent fumigant activity against Sitophilus oryzae and Tribolium castaneum, two most common insects affecting stored products. This ability makes it a promising candidate for the development of biocontrol agents against these insects.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

151.7 °F - closed cup

Flash Point(C)

66.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition of the leaf and flower essential oils of Tunisian Lavandula dentata L.(Lamiaceae).
Touati B, et al.
Chemistry and Biodiversity, 8(8), 1560-1569 (2011)
Fumigant and contact toxicities of monoterpenes to Sitophilus oryzae (L.) and Tribolium castaneum (Herbst) and their inhibitory effects on acetylcholinesterase activity.
Abdelgaleil SA, et al.
Journal of Chemical Ecology, 35(5), 518-525 (2009)
Giovanna Longhi et al.
The journal of physical chemistry. A, 110(15), 4958-4968 (2006-04-14)
We report and discuss the infrared (IR) vibrational circular dichroism (VCD) spectra of the enantiomeric pairs of the olefin derivatives of fenchone (1,3,3-trimethyl-2-methylenebicyclo[2.2.1]heptane) and camphor (1,7,7-trimethyl-2-methylenebicyclo[2.2.1]heptane), respectively, together with those of the parent molecules. The VCD spectra were taken in
I M Isa et al.
Talanta, 87, 230-234 (2011-11-22)
A new cobalt(II) ion selective electrode based on palladium(II) dichloro acetylthiophene fenchone azine(I) has been developed. The best membrane composition is found to be 10:60:10:21.1 (I)/PVC/NaTPB/DOP (w/w). The electrode exhibits a Nerstian response in the range of 1.0 × 10(-1)-1.0
Olga Tzakou et al.
Natural product communications, 4(8), 1103-1106 (2009-09-23)
The essential oils from leaves and inflorescences of L. cariensis Boiss. and L. stoechas L. subsp. stoechas collected in Greece were analyzed by GC and GC/MS. In the inflorescences and leaves essential oils of L. cariensis the most abundant metabolite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service