Skip to Content
Merck
All Photos(1)

Key Documents

W332208

Sigma-Aldrich

Thiamine hydrochloride

≥98%, FCC, FG

Synonym(s):

Aneurine hydrochloride, Vitamin B1 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27
FEMA Number:
3322
Beilstein:
3851771
EC Number:
Council of Europe no.:
4226
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.027
NACRES:
NA.21
Organoleptic:
sour
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC

Assay

≥98%

form

powder or crystals

color

white to yellow-white

mp

250 °C (dec.) (lit.)

solubility

H2O: 1 g/10 mL, clear, colorless

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sour

SMILES string

CC1=NC(N)=C(C[N+]2=CSC(CCO)=C2C)C=N1.Cl.[Cl-]

InChI

1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1

InChI key

DPJRMOMPQZCRJU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Thiamine hydrochloride is a water-soluble vitamin generally used as a nutrient supplement.[1] When it undergoes heat-degradation, it may form volatile flavor compounds such as hydrogen sulfide, 2-methylthiopehene, 2-methylfuran, and 4,5-dihydro-2-methylthiophene.[2]

Application


  • Construction of hyaluronic acid-functionalized magnolol nanoparticles for ulcerative colitis treatment.: This innovative study explores the synthesis of nanoparticle systems using hyaluronic acid-functionalized magnolol, highlighting thiamine hydrochloride′s role in enhancing therapeutic efficacy and stability in gastrointestinal treatments. This research underlines the biochemical application of thiamine hydrochloride in developing advanced drug delivery systems (Li Y et al., 2024).

  • Constructing Highly Stable Zinc Metal Anodes via Induced Zn(002) Growth.: Investigates the electrochemical stability of zinc anodes, with thiamine hydrochloride playing a critical role in modulating the electrochemical properties and enhancing the anode′s performance, showcasing its potential in next-generation battery technologies (Hu S et al., 2024).

  • Subclinical thiamine deficiency results in failed reproduction in Arctic foxes.: This study identifies the pivotal role of thiamine hydrochloride in preventing reproductive failures linked to nutritional deficiencies in wildlife, extending its application beyond human health to include environmental and conservation aspects (Jackson C et al., 2024).

  • Density Functional Theory and Density Functional Tight Binding Studies of Thiamine Hydrochloride Hydrates.: Highlights computational studies on the molecular structure and stability of thiamine hydrochloride, providing crucial insights into its physicochemical properties and interactions at the molecular level, critical for pharmaceutical formulations (Napiórkowska E et al., 2023).

  • Effects of Nutrients on the Growth of and Free Exopolysaccharide Biosynthesis by Streptococcus thermophilus 937 in a Chemically Defined Medium.: Examines the influence of thiamine hydrochloride among other nutrients on the microbial synthesis processes, underlining its essential role in microbial nutrition and metabolic engineering (Wa Y et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of HPLC method for determination of thiamine hydrochloride, riboflavin, nicotinamide, and pyridoxine hydrochloride in syrup preparation
Yantih N, et al.
Canadian Journal on Scientific and Industrial Research, 2(7), 269-278 (2011)
Spectrophotometric determination of ascorbic acid and thiamine hydrochloride in pharmaceutical products using derivative spectrophotometry
Abdel-Hamid ME, et al.
Analyst, 110(7), 831-835 (1985)
Encyclopedia of Food and Color Additives, 1, 2777-2777 null
Volatile flavor compounds produced by heat degradation of thiamine (vitamin B1).
Arnold RG, et al.
Journal of Agricultural and Food Chemistry, 17(2), 390-392 (1969)
High-performance liquid chromatography method for the simultaneous determination of thiamine hydrochloride, pyridoxine hydrochloride and cyanocobalamin in pharmaceutical formulations using coulometric electrochemical and ultraviolet detection
Marszall LM, et al.
Journal of Chromatography A, 1094(1-2), 91-98 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service