W398705
4-Hydroxybenzyl alcohol
≥98%, FG
Synonym(s):
NSC 227926
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
FEMA Number:
3987
Beilstein:
1858967
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.165
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens
Recommended Products
biological source
synthetic
grade
FG
reg. compliance
EU Regulation 1334/2008 & 178/2002
FDA
Assay
≥98%
mp
114-122 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
fruity; sweet
SMILES string
OCc1ccc(O)cc1
InChI
1S/C7H8O2/c8-5-6-1-3-7(9)4-2-6/h1-4,8-9H,5H2
InChI key
BVJSUAQZOZWCKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lei-Chwen Lin et al.
Journal of pharmaceutical and biomedical analysis, 48(3), 909-917 (2008-09-02)
Gastrodin is a pharmacologically active substance isolated from Gastrodia elata Blume with sedation, anti-convulsion and anti-epilepsy activities. A rapid and sensitive liquid chromatography technique coupled to tandem mass spectrometry (LC-MS/MS) system was developed to determine gastrodin and its metabolite p-hydroxybenzyl
Shan-Shan Yu et al.
Brain research, 1308, 167-175 (2009-10-28)
4-hydroxybenzyl alcohol (4-HBA), one of the major active phenolic constituents of Gastrodia elata Blume, a very important traditional Chinese medicinal herb, has been shown to be an effective agent against the central and peripheral nervous disorders. In this study, we
Eun-Ju Lim et al.
The Journal of pharmacy and pharmacology, 59(9), 1235-1240 (2007-09-22)
4-Hydroxybenzyl alcohol (HBA), one of the well-known phenolic compounds in diverse plants, displayed a significant inhibition in the chick chorioallantoic membrane (CAM) angiogenesis assay. HBA was shown to contain an anti-inflammatory activity in carrageenan-induced air pouch model in rats and
M W Laschke et al.
British journal of pharmacology, 163(4), 835-844 (2011-03-04)
4-Hydroxybenzyl alcohol (HBA) is a phenolic plant compound, which has been shown to influence many cellular mechanisms. In the present study, we analysed in vitro and in vivo the anti-angiogenic actions of this pleiotropic agent. Migration and protein expression of
Byung-Wook Kim et al.
Journal of ethnopharmacology, 139(2), 549-557 (2011-12-14)
Gastrodia elata Blume (Orchidaceae, GE) a traditional plant in Oriental countries is known for its enormous benefits to treat headaches, dizziness, vertigo and convulsive illnesses. In the present study, the ethnopharmacological role of GE in neuroinflammation mediated by activated microglia
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service