Skip to Content
Merck
All Photos(3)

Key Documents

H20806

Sigma-Aldrich

4-Hydroxybenzyl alcohol

99%

Synonym(s):

NSC 227926

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
1858967
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder or crystals

reaction suitability

reagent type: cross-linking reagent

mp

114-122 °C (lit.)

SMILES string

OCc1ccc(O)cc1

InChI

1S/C7H8O2/c8-5-6-1-3-7(9)4-2-6/h1-4,8-9H,5H2

InChI key

BVJSUAQZOZWCKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxybenzyl alcohol (HBA) is a phenolic compound found in Gastrodia elata. It is an antioxidant and anti-asthmatic agent.HBA can also be used as a precursor to synthesize hydrogen peroxide-responsive copolyoxalate nanoparticles and polymers.

Application

4-Hydroxybenzyl alcohol can be used as a precursor for the synthesis of:
  • HBA-incorporated copolyoxalate (HPOX) nanoparticles as potential drug delivery system.
  • Poly(4-hydroxybenzyl alcohol) by enzyme catalyzed reaction.
  • 4-Hydroxybenzyl imidazole derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen peroxide-responsive copolyoxalate nanoparticles for detection and therapy of ischemia-reperfusion injury
D Lee, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 172, 1102-1110 (2013)
Synthesis and biochemical evaluation of a range of sulfonated derivatives of 4-hydroxybenzyl imidazole as highly potent inhibitors of rat testicular 17α-hydroxylase/17, 20-lyase (P-45017α).
Ahmed S, et al.
Bioorganic & Medicinal Chemistry Letters, 19(16), 4698-4701 (2009)
Enzyme mediated oxidative polymerization of 4-hydroxybenzyl alcohol for optical applications.
Kumar V, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 39(10), 1183-1193 (2002)
Changjun Park et al.
Polymers, 12(2) (2020-02-07)
Here we report the dual light- and thermo-responsive behavior of well-defined rod-coil block copolymers composed of an azobenzene unit, 2-(2-methoxyethoxy)ethyl methacrylate (MEO2MA) and oligo(ethylene glycol) methacrylate (OEGMA). Azobenzene-containing rigid rod blocks prepared by chain growth condensation polymerization of the azobenzene
Byung-Wook Kim et al.
Journal of ethnopharmacology, 139(2), 549-557 (2011-12-14)
Gastrodia elata Blume (Orchidaceae, GE) a traditional plant in Oriental countries is known for its enormous benefits to treat headaches, dizziness, vertigo and convulsive illnesses. In the present study, the ethnopharmacological role of GE in neuroinflammation mediated by activated microglia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service