Skip to Content
Merck
All Photos(1)

Key Documents

W394505

Sigma-Aldrich

p-Anisic acid

≥99%, FG

Synonym(s):

4-Methoxybenzoic acid, p-Methoxybenzoic acid, Draconic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
FEMA Number:
3945
Beilstein:
508910
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.071
NACRES:
NA.21
Organoleptic:
animal
grade:
FG
Fragrance grade
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥99%

mp

182-185 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

animal

SMILES string

COc1ccc(cc1)C(O)=O

InChI

1S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ZEYHEAKUIGZSGI-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stephen G Bell et al.
Dalton transactions (Cambridge, England : 2003), 41(28), 8703-8714 (2012-06-15)
The crystal structures of the 4-methoxybenzoate bound forms of cytochrome P450 enzymes CYP199A2 and CYP199A4 from the Rhodopseudomonas palustris strains CGA009 and HaA2 have been solved. The structures of these two enzymes, which share 86% sequence identity, are very similar
A Hage et al.
Applied microbiology and biotechnology, 52(6), 834-838 (2000-01-05)
Ligninolytic basidiomycetes were screened for their ability to reduce aryl acids to the corresponding aldehydes and alcohols. Seven fungal strains converted p-anisic acid in high molar yields to the reduced products. The white-rot fungus Bjerkandera sp. strain BOS55 was one
M Wessels et al.
Journal of natural products, 64(12), 1556-1558 (2002-01-05)
From the hydrophilic extract of the ascidian Polycarpa aurata three new compounds, N-(4-methoxybenzoyl)-N'-methylguanidine (1), butyl 2-(4-methoxyphenyl)-2-oxoacetate (2), and 2-(4-methoxyphenyl)-N-methyl-2-oxoacetamide (3), together with the known compounds methyl 2-(4-methoxyphenyl)-2-oxoacetate (4) and 4-methoxybenzoic acid were isolated. The structures of all isolates were determined
U Karlson et al.
Journal of bacteriology, 175(5), 1467-1474 (1993-03-01)
A red-pigmented coryneform bacterium, identified as Rhodococcus rhodochrous strain 116, that grew on 2-ethoxyphenol and 4-methoxybenzoate as sole carbon and energy sources was isolated. Phylogenetic analysis based on the 16S rDNA sequences indicates that the strain clusters more closely to
K Sekar et al.
Acta crystallographica. Section D, Biological crystallography, 62(Pt 4), 392-397 (2006-03-23)
The lipolytic enzyme phospholipase A2 plays a crucial role in lipid metabolism and catalyzes hydrolysis of the fatty-acid ester bond at the sn-2 position of phospholipids. Here, the crystal structure (1.7 A resolution) of the triple mutant (K53,56,121M) of bovine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service