Skip to Content
Merck
All Photos(1)

Key Documents

W242000

Sigma-Aldrich

Ethyl p-anisate

≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2C2H5
CAS Number:
Molecular Weight:
180.20
FEMA Number:
2420
EC Number:
Council of Europe no.:
249
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.714
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
biological source:
synthetic

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.524 (lit.)

bp

263 °C (lit.)

mp

7-8 °C (lit.)

density

1.103 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

fragrance allergen

no known allergens

Organoleptic

anise; fruity

SMILES string

CCOC(=O)c1ccc(OC)cc1

InChI

1S/C10H12O3/c1-3-13-10(11)8-4-6-9(12-2)7-5-8/h4-7H,3H2,1-2H3

InChI key

FHUODBDRWMIBQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service