Skip to Content
Merck
All Photos(1)

Key Documents

W393899

Sigma-Aldrich

Nopol

≥98%, FG

Synonym(s):

10-Hydroxymethylene-2-pinene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
FEMA Number:
3938
UNSPSC Code:
12164502
Flavis number:
02.141
NACRES:
NA.05

grade

FG
Halal
Kosher

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsamic; citrus; pine; sweet

SMILES string

OCCC1=CCC2CC1C2(C)C

InChI

1S/C11H18O/c1-11(2)9-4-3-8(5-6-12)10(11)7-9/h3,9-10,12H,4-7H2,1-2H3

InChI key

ROKSAUSPJGWCSM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service