Skip to Content
Merck
All Photos(1)

Key Documents

W352316

Sigma-Aldrich

Pyrrolidine

FG

Synonym(s):

Tetrahydropyrrole, Tetramethyleneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9N
CAS Number:
Molecular Weight:
71.12
FEMA Number:
3523
Beilstein:
102395
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.064
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor density

2.45 (vs air)

vapor pressure

128 mmHg ( 39 °C)
49 mmHg ( 20 °C)

autoignition temp.

653 °F

expl. lim.

10.6 %

refractive index

n20/D 1.443 (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy

SMILES string

C1CCNC1

InChI

1S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2

InChI key

RWRDLPDLKQPQOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph T Paletta et al.
Organic letters, 14(20), 5322-5325 (2012-10-12)
A series of sterically shielded pyrrolidine nitroxides were synthesized, and their reduction by ascorbate (vitamin C) indicate that nitroxide 3, a tetraethyl derivative of 3-carboxy-PROXYL, is reduced at the slowest rate among known nitroxides, i.e., at a 60-fold slower rate
Adele Faulkner et al.
Chemical communications (Cambridge, England), 49(15), 1521-1523 (2013-01-17)
We report efficient Pd-catalyzed cyclizations of oxime esters with 1,1-disubstituted alkenes as the basis of a general entry to α,α-disubstituted pyrrolidine derivatives. We also demonstrate that catalytic asymmetric variants of this chemistry are feasible by employing a suitable chiral ligand.
Pieter Van der Veken et al.
Journal of medicinal chemistry, 55(22), 9856-9867 (2012-11-06)
We have investigated the effect of regiospecifically introducing substituents in the P2 part of the typical dipeptide derived basic structure of PREP inhibitors. This hitherto unexplored modification type can be used to improve target affinity, selectivity, and physicochemical parameters in
Boyu Zhang et al.
Angewandte Chemie (International ed. in English), 51(52), 13159-13162 (2012-11-20)
Drop it! A highly enantioselective catalytic cascade reaction of α-ketoacids and aldehydes is achieved using the title catalyst and water as the solvent. Fluorescence imaging shows that the catalyst is mainly distributed on the surface of emulsion droplets. Optically active
Igor A Kirilyuk et al.
The Journal of organic chemistry, 77(18), 8016-8027 (2012-08-25)
The nitroxides of 7-azadispiro[5.1.5.2]pentadecane and 7-azadispiro[5.1.5.2]pentadeca-14-ene series have been prepared, including thiol-specific methane thiosulfonate spin label for site-directed spin labeling. The effect of spirocyclohexane moieties on chemical and spectral properties has been studied. The obtained temperature dependencies of electron spin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service