Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrrolidine
Synonym(s):
Tetrahydropyrrole, Tetramethyleneimine, Pyrrolidine
Empirical Formula (Hill Notation):
C
4
H
9
N
CAS Number:
123-75-1
Molecular Weight:
71.12
EC Number:
204-648-7
Products
Product Category
building blocks (3)
aroma chemicals (1)
Brand
Sigma-Aldrich (4)
Biological Source
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Food Grade (1)
Halal (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
P73803
Pyrrolidine,
99%
Expand
View Pricing
394238
Pyrrolidine,
≥99.5%, purified by redistillation
Expand
View Pricing
8.07494
Pyrrolidine,
for synthesis
Expand
View Pricing
W352316
Pyrrolidine,
FG
Expand
View Pricing
83240
Pyrrolidine,
≥99.0%
Expand
View Pricing