Skip to Content
Merck
All Photos(1)

Documents

W329118

Sigma-Aldrich

4-Hydroxybutanoic acid lactone

≥98%, FCC, FG

Synonym(s):

γ-Butyrolactone, γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
FEMA Number:
3291
Beilstein:
105248
EC Number:
Council of Europe no.:
615
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.006
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA

vapor density

3 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

Assay

≥98%

autoignition temp.

851 °F

expl. lim.

16 %

refractive index

n20/D 1.436 (lit.)

bp

204-205 °C (lit.)

mp

−45 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; creamy; fatty; oily

SMILES string

O=C1CCCO1

InChI

1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2

InChI key

YEJRWHAVMIAJKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • RIFM fragrance ingredient safety assessment, 4-hydroxybutanoic acid lactone, CAS Registry Number 96-48-0.: A comprehensive safety assessment of 4-hydroxybutanoic acid lactone as a fragrance ingredient, evaluating its toxicological profile and safe usage levels (Api et al., 2019).

Biochem/physiol Actions

Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michelle Wood et al.
Journal of chromatography. A, 1056(1-2), 83-90 (2004-12-15)
We have developed a rapid method that enables the simultaneous analysis of gamma-hydroxybutyrate (GHB) and its precursors, i.e. gamma-butyrolactone (GBL) and 1,4-butanediol (1,4-BD) in urine. The method comprised a simple dilution of the urine sample, followed by liquid chromatography-tandem mass
Zheng-Yuan Su et al.
Chemical research in toxicology, 26(3), 477-485 (2013-02-28)
Cancer development has been linked to epigenetic modifications of cancer oncogenes and tumor suppressor genes; in advanced metastatic cancers, severe epigenetic modifications are present. We previously demonstrated that the progression of prostate tumors in TRAMP mice is associated with methylation
Christian Westendorf et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), 3853-3858 (2013-02-23)
The rapid reorganization of the actin cytoskeleton in response to external stimuli is an essential property of many motile eukaryotic cells. Here, we report evidence that the actin machinery of chemotactic Dictyostelium cells operates close to an oscillatory instability. When
Bo Xue et al.
Biochemistry, 52(13), 2359-2370 (2013-03-07)
The in vitro evolution and engineering of quorum-quenching lactonases with enhanced reactivities was achieved using a thermostable GKL enzyme as a template, yielding the E101G/R230C GKL mutant with increased catalytic activity and a broadened substrate range [Chow, J. Y., Xue
Jagan N Thupari et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(14), 9498-9502 (2002-06-13)
C75, a known inhibitor of fatty acid synthase is postulated to cause significant weight loss through decreased hypothalamic neuropeptide Y (NPY) production. Peripherally, C75, an alpha-methylene-gamma-butyrolactone, reduces adipose tissue and fatty liver, despite high levels of malonyl-CoA. To investigate this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service