Skip to Content
Merck
All Photos(1)

Key Documents

W308005

Sigma-Aldrich

Tributyl 2-acetylcitrate

≥98%, FG

Synonym(s):

Acetyl tributyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C[CO2(CH2)3CH3][CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
402.48
FEMA Number:
3080
Beilstein:
2303316
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.511
NACRES:
NA.21
grade:
FG
Fragrance grade
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

vapor pressure

0.26 psi ( 20 °C)
0.8 psi ( 55 °C)

Assay

≥98%

refractive index

n20/D 1.443 (lit.)

density

1.05 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

herbaceous; sweet; wine-like

SMILES string

CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC

InChI

1S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3

InChI key

QZCLKYGREBVARF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effect of Starch Plasticization on Morphological, Mechanical, Crystalline, Thermal, and Optical Behavior of Poly(butylene adipate-co-terephthalate)/Thermoplastic Starch Composite Films.: This study investigates the influence of starch plasticization on various properties of poly(butylene adipate-co-terephthalate) composite films. Tributyl 2-acetylcitrate is evaluated as a plasticizer to enhance the material characteristics, with findings indicating improved mechanical and thermal performance (He et al., 2024, doi:10.3390/polym16030326).

  • Toxicological assessment of phthalates and their alternatives using human keratinocytes.: This research examines the toxicological effects of various phthalates and alternative plasticizers, including Tributyl 2-acetylcitrate, on human keratinocytes. The study provides insights into the safety and potential health impacts of these compounds (Kim et al., 2019, doi:10.1016/j.envres.2019.05.007).

  • Developmental toxicity of dibutyl phthalate and citrate ester plasticizers in Xenopus laevis embryos.: This study explores the developmental toxicity of dibutyl phthalate and citrate ester plasticizers, including Tributyl 2-acetylcitrate, in amphibian embryos. The research highlights potential developmental risks associated with these compounds (Xu et al., 2018, doi:10.1016/j.chemosphere.2018.04.077).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

422.6 °F - closed cup

Flash Point(C)

217 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Pay et al.
Lasers in surgery and medicine, 24(1), 48-54 (1999-02-26)
Many laser centres utilise various transparent membranes during treatment sessions with Q-switched lasers to prevent or reduce tissue splatter, thereby reducing the aerosolised biohazard of cellular debris to operator and laser. We performed a series of experiments with the Q-Switched
P D Zygoura et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 28(5), 686-694 (2011-03-15)
Migration levels of acetyl tributyl citrate (ATBC) plasticiser from polyvinyl chloride (PVC) film into the European Union specified aqueous food simulants (distilled water, 3% w/v acetic acid and 10% v/v ethanol) were monitored as a function of time. Migration testing
C E Dörfer et al.
Journal of clinical periodontology, 27(3), 162-168 (2000-04-01)
The comparison of the effects of guided-tissue regeneration (GTR) using 2 different biodegradable barriers (polylactide acetyltributyl citrate; polydioxanon) in 3- and 2-wall intrabony defects. The polydioxanon barrier is an experimental membrane for GTR therapy that consists of an continuous occlusive
Acetyl tributyl citrate and dibutyl sebacate inhibit the growth of cultured mammalian cells.
K Mochida et al.
Bulletin of environmental contamination and toxicology, 56(4), 635-637 (1996-04-01)
Guang Hong et al.
Dental materials journal, 29(4), 374-380 (2010-07-09)
The purpose of this study was to determine the effect of PMMA polymer on dynamic viscoelasticity and plasticizer leachability of PEMA-based tissue conditioners. One PEMA polymer and one PMMA polymer were used in powder form with four formulations. The combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service