Skip to Content
Merck
All Photos(3)

Key Documents

W285811

Sigma-Aldrich

Phenethyl alcohol

greener alternative

natural, ≥99%, FCC, FG

Synonym(s):

2-Phenylethanol, β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
FEMA Number:
2858
Beilstein:
1905732
EC Number:
Council of Europe no.:
68
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.019
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

4.21 (vs air)

vapor pressure

1 mmHg ( 58 °C)

Assay

≥99%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.5317 (lit.)

bp

219-221 °C/750 mmHg (lit.)

mp

−27 °C (lit.)

density

1.020 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

honey; floral; rose

SMILES string

OCCc1ccccc1

InChI

1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

WRMNZCZEMHIOCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phenethyl alcohol, an aromatic alcohol with rose-like odor, is commonly used as a food flavoring and fragrance ingredient. It is the main flavor volatile of tomato and blue cheese.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased product, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

recommended

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of 2?Phenylethyl Alcohol by Kluyveromyces marxianus.
Fabre CE, et al.
Biotechnology Progress, 14(2), 270-274 (1998)
Production of 2-phenylethanol, a flavor ingredient, by Pichia fermentans L-5 under various culture conditions.
Huang CJ, et al.
Food Research International, 34(4), 277-282 (2001)
Quantitation, evaluation, and effect of certain microorganisms on flavor components of blue cheese.
Anderson DF & Day EA.
Journal of Agricultural and Food Chemistry, 14(3), 241-245 (1966)
Sara Invitto et al.
Respiratory physiology & neurobiology, 259, 37-44 (2018-07-15)
Obstructive Sleep Apnea Syndrome (OSA) is characterized by snoring associated with repeated apnea and/or obstructive hypopnea. The nasal airways of OSA patients, measured via acoustic rhinometry, could be significantly narrower than healthy subjects and this reduced nasal structure can impair
Sara Invitto et al.
Frontiers in aging neuroscience, 10, 272-272 (2018-10-03)
Olfactory impairment is present in up to 90% of patients with Alzheimer's disease (AD) and is present in certain cases of mild cognitive impairment (MCI), a transient phase between normal aging and dementia. Subjects affected by MCI have a higher

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service