Skip to Content
Merck
All Photos(1)

Key Documents

W280518

Sigma-Aldrich

1-Octen-3-ol

≥98%, FCC, FG

Synonym(s):

Pentyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH=CH2
CAS Number:
Molecular Weight:
128.21
FEMA Number:
2805
Beilstein:
1744110
EC Number:
Council of Europe no.:
72
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.023
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.437 (lit.)

bp

84-85 °C/25 mmHg (lit.)

density

0.837 g/mL at 20 °C
0.83 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; mushroom; oily; earthy

SMILES string

CCCCCC(O)C=C

InChI

1S/C8H16O/c1-3-5-6-7-8(9)4-2/h4,8-9H,2-3,5-7H2,1H3

InChI key

VSMOENVRRABVKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Octen-3-ol occurs naturally in mushrooms. It is the most prominent odor volatile produced by fungi such as Aspergillus, Penicillium and Fusarium species. 1-Octen-3-ol is also formed in major quantity when soybeans are soaked in water as a pretreatment before soymilk production. It is also a potent mosquito attractant. 1-Octen-3-ol is the major volatile compound formed during autoxidation of linoleic acid.

Biochem/physiol Actions

Taste at 0.01-0.20 ppm

Other Notes

Natural occurrence: Clover, cooked artichoke, fish, mushroom, peas, soybeans, wheat bread, tea, and beef.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The formation of 1-octen-3-ol in soybeans during soaking.
Badenhop AF & Wilkens WF.
Journal of the American Oil Chemists' Society, 46(3), 179-182 (1969)
Carbon dioxide and 1-octen-3-ol as mosquito attractants.
Takken W & Kline DL.
Journal of the American Mosquito Control Association, 5(3), 311-316 (1989)
Studies on mushroom flavours 3. Some flavour compounds in fresh, canned and dried edible mushrooms.
Dijkstra FY.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 160(4), 401-405 (1976)
Gabriel Z Laporta et al.
Memorias do Instituto Oswaldo Cruz, 106(3), 279-284 (2011-06-10)
Studies have shown that both carbon dioxide (CO₂) and octenol (1-octen-3-ol) are effective attractants for mosquitoes. The objective of the present study was to evaluate the attractiveness of 1-octen-3-ol and CO₂ for diurnal mosquitoes in the southeastern Atlantic forest. A
L E Harrup et al.
Journal of medical entomology, 49(1), 112-121 (2012-02-09)
The host kairomones carbon dioxide (CO2) and 1-octen-3-ol elicit a host seeking response in a wide range of haematophagous Diptera. This study investigates the response of Culicoides biting midges (Diptera: Ceratopogonidae) to these cues using field-based experiments at two sites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service