Skip to Content
Merck
All Photos(1)

Key Documents

W249106

Sigma-Aldrich

Furfuryl alcohol

≥97%, FG

Synonym(s):

2-(Hydroxymethyl)furan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
FEMA Number:
2491
Beilstein:
106291
EC Number:
Council of Europe no.:
2023
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.019
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 175.105

vapor density

3.4 (vs air)

vapor pressure

0.5 mmHg ( 20 °C)
1 mmHg ( 32 °C)
5.5 mmHg ( 55 °C)

Assay

≥97%

autoignition temp.

915 °F

expl. lim.

16.3 %

refractive index

n20/D 1.486 (lit.)

bp

170 °C (lit.)

mp

−29 °C (lit.)

density

1.135 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

bread; caramel; brown; sweet

SMILES string

OCc1ccco1

InChI

1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2

InChI key

XPFVYQJUAUNWIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Furfuryl alcohol is a volatile flavor compound identified in nonfat dry milk[1], rice cakes[2] and honey.[3]

Application


  • Achmatowicz Rearrangement-Inspired Development of Green Chemistry, Organic Methodology, and Total Synthesis of Natural Products.: This study discusses innovative organic synthesis strategies inspired by the Achmatowicz rearrangement, focusing on green chemistry and the synthesis of natural products, which could include applications of furfuryl alcohol as a precursor or reagent (Liang et al., 2022).

  • Triplet Photochemistry of Dissolved Black Carbon and Its Effects on the Photochemical Formation of Reactive Oxygen Species.: This paper examines the triplet-state photochemistry of dissolved black carbon, which may implicate the photochemical behaviors of furfuryl alcohol in environmental matrices (Wang et al., 2020).

  • Production of Singlet Oxygen ((1)O(2)) during the Photochemistry of Aqueous Pyruvic Acid: The Effects of pH and Photon Flux under Steady-State O(2)(aq) Concentration.: Investigates singlet oxygen production in aqueous pyruvic acid, potentially relevant to understanding the photochemical properties of furfuryl alcohol (Eugene and Guzman, 2019).

  • Triplet-State Photochemistry of Dissolved Organic Matter: Triplet-State Energy Distribution and Surface Electric Charge Conditions.: Discusses the triplet-state energy of dissolved organic matter, which could relate to the behavior of furfuryl alcohol in various environmental conditions (Zhou et al., 2019).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nose, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimized Likens-Nickerson methodology for quantifying honey flavors.
Bouseta A & Collin S.
Journal of Agricultural and Food Chemistry, 43(7), 1890-1897 (1995)
Samuel D Snow et al.
Environmental science & technology, 46(24), 13227-13234 (2012-11-16)
The vast range of C₆₀ derivatives makes it difficult to assess the potential environmental impact of this class of materials, while past environmental studies mostly focused only on pristine C₆₀. Central to derivatized C₆₀'s potential to negatively impact (micro)biological receptors
Zehui Zhang et al.
ChemSusChem, 4(1), 112-118 (2011-01-13)
A clean, facile, and environment-friendly catalytic method has been developed for the conversion of furfuryl alcohol into alkyl levulinates making use of the novel solid catalyst methylimidazolebutylsulfate phosphotungstate ([MIMBS]₃PW₁₂O₄₀). The solid catalyst is an organic-inorganic hybrid material, which consists of
Bernhard H Monien et al.
Carcinogenesis, 32(10), 1533-1539 (2011-07-07)
Furfuryl alcohol, formed by acid- and heat-induced dehydration from pentoses, is found in many foodstuffs. It induced renal tubule neoplasms in male B6C3F1 mice and nasal neoplasms in male F344/N rats in a study of the National Toxicology Program (NTP).
Highly selective decarbonylation of 5-(hydroxymethyl)furfural in the presence of compressed carbon dioxide.
Frank M A Geilen et al.
Angewandte Chemie (International ed. in English), 50(30), 6831-6834 (2011-06-11)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service