Skip to Content
Merck
All Photos(1)

Key Documents

44519

Supelco

Dowex 50W X8 Ion Exchange Resin

hydrogen form, 200-400 mesh, strongly acidic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

Product Name

Dowex 50W X8, hydrogen form, strongly acidic, 200-400 mesh

description

cross-linkage 8%

Quality Level

form

beads

moisture

50-58%

technique(s)

LPLC: suitable

loss

~55% loss on drying, 110 °C

matrix

styrene divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

200-400 mesh

capacity

1.1 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dowex 50WX8 hydrogen form is used as stripper resin and can be regenerated by 0.25N sulphuric acid when exhausted. Dowex 50WX8 hydrogen form is used as catalyst. It is used in deionizing supernatant fraction samples for paper chromatography.

Application

Dowex 50WX8 hydrogen form may be used in separation medium for TLC, paper chromatography and LPLC.

Legal Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition and ultrastructure of wild-type and white mutantAspergillus nidulans conidial walls.
Claverie-Martin, Felix, Maria R. Diaz-Torres, and Michael J. Geoghegan.
Current Microbiology, 16, 281-287 (1988)
Novel ion exchange chromatographic method using conductimetric detection.
Small, Hamish, Timothy S. Stevens, and William C. Bauman.
Analytical Chemistry, 47, 1801-1809 (1975)
Hydrogen generation from hydrolysis of sodium borohydride using Ni-Ru nanocomposite as catalysts.
Liu, Cheng-Hong, et al.
International Journal of Hydrogen Energy, 34, 2153-2163 (2009)
Jianghong Yan et al.
Acta biochimica et biophysica Sinica, 52(12), 1360-1372 (2020-11-25)
Inducible nitric oxide synthase (iNOS) plays critical roles in the inflammatory response and host defense. Previous research on iNOS regulation mainly focused on its gene expression level, and much less is known about the regulation of iNOS function by N-glycosylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service