Skip to Content
Merck
All Photos(1)

Key Documents

V1503

Sigma-Aldrich

Vinyl acetate

contains 3-20 ppm hydroquinone as inhibitor, ≥99%

Synonym(s):

VAc, Acetoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH=CH2
CAS Number:
Molecular Weight:
86.09
Beilstein:
1209327
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

Quality Level

vapor pressure

88 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

801 °F

contains

3-20 ppm hydroquinone as inhibitor

expl. lim.

13.4 %

refractive index

n20/D 1.395 (lit.)

bp

72-73 °C (lit.)

mp

−93 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)OC=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Vinyl acetate has been used as an acyl donor for enzyme-catalyzed acylation reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipases efficiently stearate and cutinases acetylate the surface of arabinoxylan films.
Stepan AM
Journal of Biotechnology, 167(1), 16-23 (2013)
María Laura Lastra et al.
Colloids and surfaces. B, Biointerfaces, 196, 111322-111322 (2020-08-26)
Recent studies have shown the relevance of growing mesenchymal stem cells (MSCs) in three-dimensional environments with respect to the monolayer cell culture on an adherent substrate. In this sense, macroporous scaffolds and hydrogels have been used as three-dimensional (3D) supports.
Kinetic resolution of 4-chloro-2-(1-hydroxyalkyl) pyridines using Pseudomonas cepacia lipase.
Busto E
Nature Protocols, 1(4), 2061-2061 (2006)
Nhan Thanh Thien Huynh et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-22)
5-Acetoxymethyl-2-furfural (AMF) was prepared from D-fructose via 1,6-diacetylfructose (DAF) through a simple two-step reaction pathway. Immobilized enzyme (Novozym 435) was found to be the best enzymatic catalyst for the trans-esterification step (yielding 94.6% DAF). In the dehydration step, while soluble
Ping Xu et al.
Polymers, 11(2) (2019-04-10)
Hyperbranched polycaprolactone with controlled structure was synthesized by reversible addition-fragmentation chain transfer radical ring-opening polymerization along with self-condensed vinyl polymerization (SCVP) of 2-methylene-1,3-dioxepane (MDO). Vinyl 2-[(ethoxycarbonothioyl) sulfanyl] propanoate (ECTVP) was used as polymerizable chain transfer agent. Living polymerization behavior was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service