Skip to Content
Merck
All Photos(1)

Key Documents

U2008

Sigma-Aldrich

10-Undecen-1-ol

98%

Synonym(s):

11-Hydroxy-1-undecene, NSC 66157, Undecylenyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)9OH
CAS Number:
Molecular Weight:
170.29
Beilstein:
1753225
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.448-1.453
n20/D 1.45 (lit.)

bp

132-133 °C/15 mmHg (lit.)
245-248 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCCCCC=C

InChI

1S/C11H22O/c1-2-3-4-5-6-7-8-9-10-11-12/h2,12H,1,3-11H2

InChI key

GIEMHYCMBGELGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service