Skip to Content
Merck
All Photos(2)

Key Documents

T81809

Sigma-Aldrich

Triphenylantimony(III)

99%

Synonym(s):

Antimony(III)triphenyl, Triphenylstibine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3Sb
CAS Number:
Molecular Weight:
353.07
Beilstein:
3608108
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

solid

reaction suitability

core: antimony
reagent type: catalyst

bp

377 °C (lit.)

mp

52-54 °C (lit.)

SMILES string

c1ccc(cc1)[Sb](c2ccccc2)c3ccccc3

InChI

1S/3C6H5.Sb/c3*1-2-4-6-5-3-1;/h3*1-5H;

InChI key

HVYVMSPIJIWUNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 51 Antimony

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service