P49406
Piperonyl alcohol
98%
Synonym(s):
1,3-Benzodioxole-5-methanol, 3,4-(Methylenedioxy)benzyl alcohol, 3,4-(Methylenedioxy)phenylmethanol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
Beilstein:
136113
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
mp
50-54 °C (lit.)
SMILES string
OCc1ccc2OCOc2c1
InChI
1S/C8H8O3/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-3,9H,4-5H2
InChI key
BHUIUXNAPJIDOG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Daniel Schwendenwein et al.
Advanced synthesis & catalysis, 358(21), 3414-3421 (2016-12-06)
The enzymatic reduction of carboxylic acids is in its infancy with only a handful of biocatalysts available to this end. We have increased the spectrum of carboxylate-reducing enzymes (CARs) with the sequence of a fungal CAR from Neurospora crassa OR74A
S R Victor et al.
Pest management science, 57(7), 603-608 (2001-07-24)
The development of Leucoagaricus gongylophorus, the fungus cultured by the leaf-cutting ant Atta sexdens was inhibited in vitro by synthetic compounds containing the piperonyl group. In addition, worker ants that were fed daily on an artificial diet to which these
Marise M O Cabral et al.
Fitoterapia, 79(1), 59-63 (2007-10-02)
The neolignan, burchellin, a natural compound that reduces urine excretion in larvae of the bloodsucking bug, Rhodnius prolixus, a vector of Chagas' disease, is rapidly degraded in the hemolymph of the insect. The main product that accumulates in this tissue
Aaron D Gross et al.
Chemico-biological interactions, 263, 1-6 (2016-12-18)
An outbreak of the southern cattle tick, Rhipicephalus (Boophilus) microplus, (Canestrini), in the United States would have devastating consequences on the cattle industry. Tick populations have developed resistance to current acaricides, highlighting the need to identify new biochemical targets along
Metabolism of piperonal and piperonyl alcohol in the rat with special reference to the scission of the methylenedioxy group.
J Klungsøyr et al.
Acta pharmaceutica Suecica, 21(1), 67-72 (1984-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service