Skip to Content
Merck
All Photos(2)

Key Documents

758078

Sigma-Aldrich

5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoic acid

97%

Synonym(s):

5-(3,4-Methylenedioxyphenyl)-2,4-pentadienoic acid, Piperic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O4
CAS Number:
Molecular Weight:
218.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

218-223 °C

SMILES string

OC(=O)\C=C\C=C\c1ccc2OCOc2c1

InChI

1S/C12H10O4/c13-12(14)4-2-1-3-9-5-6-10-11(7-9)16-8-15-10/h1-7H,8H2,(H,13,14)/b3-1+,4-2+

InChI key

RHBGITBPARBDPH-ZPUQHVIOSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Satyendra Mishra et al.
Bioorganic & medicinal chemistry, 13(5), 1477-1486 (2005-02-09)
In the present communication different curcumin bioconjugates viz. 4,4'-di-O-glycinoyl-curcumin, 4,4'-di-O-d-alaninoyl-curcumin, 4,4'-di-O-(glycinoyl-di-N-piperoyl)-curcumin, 4,4'-di-O-piperoyl curcumin, curcumin-4,4'-di-O-beta-d-glucopyranoside, 4,4'-di-O-acetyl-curcumin along with piperoyl glycine, have been synthesised and characterised by spectra UV, (1)H NMR and elemental analysis. All the covalent bonds used are biodegradable. This
Shiv K Dubey et al.
European journal of medicinal chemistry, 43(9), 1837-1846 (2008-01-19)
The monoesters of curcumin, a symmetric diphenol with valine and glycine have been prepared by a novel solid phase synthesis and its diesters with valine, glutamic acid and demethylenated piperic acid have been prepared by solution phase method. The assessment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service