Skip to Content
Merck
All Photos(1)

Key Documents

P3265

Sigma-Aldrich

Poly(ethylene glycol)

average MN 400

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol), average mol wt 400

form

liquid

mol wt

average mol wt 400

refractive index

n/D 1.465 (20 °C)

viscosity

7.3 cSt (210 °F)

mp

4-8 °C

solubility

water: 100 mg/mL

density

1.1254 g/cm3 at 20 °C
1.0930 g/cm3 at 60 °C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is an oligomer of ethylene oxide that is majorly used as a synthetic polymer for its unique properties which include low cost, biocompatibility, and hydrophilicity. It can be used in the production of biomedical hydrogels.

Application

PEG has a variety of biomedical applications, applications in energy storage and drug delivery.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Nakayama et al.
Developmental biology, 426(2), 472-486 (2016-09-07)
We describe a novel recessive and nonlethal pigmentation mutant in Xenopus tropicalis. The mutant phenotype can be initially observed in tadpoles after stage 39/40, when mutant embryos display markedly reduced pigmentation in the retina and the trunk. By tadpole stage
Hydrogels in craniofacial tissue engineering
Biomaterials for Oral and Dental Tissue Engineering, 47-64 (2017)
Entrapment of essential oils in hydrogels for biomedical applications
Polymeric Gels, 125-141 (2018)
Biomedical hydrogels
Biomaterials, Artificial Organs and Tissue Engineering, 107-115 (2005)
Vanessa Martínez-Francés et al.
Frontiers in pharmacology, 8, 467-467 (2017-08-10)
In Valencia Region (Spain), some wild and cultivated sages are used for medicinal purposes. Among them

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service