Skip to Content
Merck
All Photos(1)

Key Documents

P1101

Sigma-Aldrich

D-Penicillamine disulfide

97%, cell analysis

Synonym(s):

3,3′-Dithiobis(2-amino-3-methylbutanoic acid), 3,3′-Dithiobis(2-amino-3-methylbutyric acid), 3,3′-Dithiobis-D-valine, S,S′-Bi(D-penicillamine), NSC 87505

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SC(CH3)2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
296.41
Beilstein:
4461521
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

product name

D-Penicillamine disulfide, 97%

Quality Level

Assay

97%

form

powder or crystals

optical activity

[α]25/D −75°, c = 1 in 1 M NaOH

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

204 °C (dec.) (lit.)

application(s)

cell analysis

SMILES string

CC(C)(SSC(C)(C)[C@@H](N)C(O)=O)[C@@H](N)C(O)=O

InChI

1S/C10H20N2O4S2/c1-9(2,5(11)7(13)14)17-18-10(3,4)6(12)8(15)16/h5-6H,11-12H2,1-4H3,(H,13,14)(H,15,16)/t5-,6-/m0/s1

InChI key

POYPKGFSZHXASD-WDSKDSINSA-N

Application

Used in pharmacological studies of:
  • Molecules dermatomyositis activity
  • Antimelanoma activity of apoptogenic carbonyl scavengers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrizio Apruzzese et al.
Annali di chimica, 94(1-2), 45-56 (2004-05-15)
D-penicillamine disulfide (PNS) shows protolytic properties and is able to form complexes with cations, because it has two aminic groups and two carboxylic groups. The four protonation constants of its deprotonated species were determined by means of electromotive force (e.m.f.)
Determination of penicillamine in encapsulated formulations by high-performance liquid chromatography.
S Biffar et al.
Journal of chromatography, 318(2), 404-407 (1985-01-18)
Amit K Galande et al.
Biopolymers, 71(5), 534-551 (2003-11-25)
A recently rediscovered reaction of base-assisted lanthionine formation has been applied to several systems of disulfide-bridged peptides. In addition to previously described nonapeptides consisting of i, i+3 cystine linkages, the reaction has now been extended to systems consisting of shorter
[Determination of D-penicillamine and its metabolites in blood and urine].
K Kyogoku et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 102(4), 322-327 (1982-04-01)
G T Yamashita et al.
Journal of chromatography, 491(2), 341-354 (1989-07-21)
Methodology is described for the simultaneous determination of D-penicillamine, penicillamine disulfide and the penicillamine-glutathione mixed disulfide, as well as glutathione and glutathione disulfide, in human plasma, erythrocytes and urine. The various thiols and disulfides are separated by reversed-phase ion-pairing liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service