Skip to Content
Merck
All Photos(3)

Key Documents

N27322

Sigma-Aldrich

4-Nitrotoluene

99%

Synonym(s):

1-Methyl-4-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
1906911
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.7 (vs air)

vapor pressure

5 mmHg ( 85 °C)

Assay

99%

form

crystals

autoignition temp.

734 °F

bp

238 °C (lit.)

mp

52-54 °C (lit.)

density

1.392 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H7NO2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3

InChI key

ZPTVNYMJQHSSEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Nitrotoluene is a nitroaromatic compound used in the synthesis of drugs, pesticides, and dyes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zinc oxide-copper sulfide semiconductor nano-heterostructure for low-level electrochemical detection of 4-nitrotoluene
Urmila C et al.
Electrochimica Acta, 447, 142160-142160 (2023)
Shuang Song et al.
Journal of hazardous materials, 144(1-2), 532-537 (2006-12-05)
p-Nitrotoluene (PNT) is a nitroaromatic compound that is hazardous to humans and is a suspected hormone disrupter. The degradation of PNT in aqueous solution by ozonation (O(3)) combined with sonolysis (US) was investigated in laboratory-scale experiments in which pH, initial
Avi Cagan et al.
The Analyst, 133(5), 585-587 (2008-04-23)
A highly selective and rapid electrochemical assay of the improvised explosive urea nitrate (UN) is reported. The method involves a short ( approximately 10 s) acid-catalyzed reaction of UN with 4-nitrotoluene (NT) followed by a rapid ( approximately 2 s)
Patricia Guerra et al.
Journal of separation science, 31(15), 2891-2898 (2008-07-31)
Ion mobility spectrometry (IMS) is routinely used in screening checkpoints for the detection of explosives and illicit drugs but it mainly relies on the capture of particles on a swab surface for the detection. Solid phase microextraction (SPME) has been
S Ramalingam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(4), 1308-1314 (2010-02-06)
In this work, the experimental and theoretical study on molecular structure and vibrational spectra of 4-nitrotoluene are studied. The FTIR and FTRaman experimental spectra of the molecule have been recorded in the range of 4000-100 cm(-1). Making use of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service