Skip to Content
Merck
All Photos(2)

Key Documents

N21588

Sigma-Aldrich

2-Nitrophenylhydrazine

97%

Synonym(s):

(2-Nitrophenyl)hydrazine, (o-Nitrophenyl)hydrazine, 1-(2-Nitrophenyl)hydrazine, 1-Hydrazino-2-nitrobenzene, 2-Hydrazino-1-nitrobenzene, o-(Hydrazino)nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NHNH2
CAS Number:
Molecular Weight:
153.14
Beilstein:
512797
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

contains

30% water as stabilizer (minimum)

mp

91-93 °C (lit.)

storage temp.

2-8°C

SMILES string

NNc1ccccc1[N+]([O-])=O

InChI

1S/C6H7N3O2/c7-8-5-3-1-2-4-6(5)9(10)11/h1-4,8H,7H2

InChI key

FRBUNLLUASHNDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vida Časaitė et al.
Applied and environmental microbiology, 86(15) (2020-05-31)
Pyridine and its derivatives constitute the majority of heterocyclic aromatic compounds that occur largely as a result of human activities and contribute to environmental pollution. It is known that they can be degraded by various bacteria in the environment; however
A I Fattom et al.
Infection and immunity, 66(10), 4588-4592 (1998-09-24)
Bacterial capsular polysaccharides (CP) are carbohydrate polymers comprised of repeating saccharide units. Several of these CP have side chains attached to their backbone structures. The side chains may include O-acetyl, phosphate, sialic acid, and other moieties. Those moieties represent the
Chikako Yomota et al.
Journal of chromatography. A, 1142(2), 231-235 (2007-01-20)
Currently, biotin is typically determined in Japan using a microbiological method. Such microbiological assays are sensitive, but they are not always highly specific and are also rather tedious and time-consuming. In the present study, RP-HPLC and LC-MS methods for the
T Takeuchi et al.
Clinica chimica acta; international journal of clinical chemistry, 205(1-2), 117-126 (1992-01-31)
An automated assay method has been developed for the measurement of serum cholinesterase activity. The samples were prepared by an automated liquid handling unit and incubated for 9.7 min at 30 degrees C, followed by automatic injection into a colorimetric
Gregory C Henderson et al.
Analytical biochemistry, 413(1), 66-68 (2011-02-18)
Previously, following derivatization to their 2-nitrophenylhydrazide (2-NPH) derivatives, fatty acid (FA) abundances have been evaluated using high-performance liquid chromatography (HPLC). Although the method was sensitive, resolution was insufficient for many of the biologically important FAs. We have developed an enhanced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service