Skip to Content
Merck
All Photos(2)

Key Documents

N11507

Sigma-Aldrich

2-Nitrobenzenesulfonyl chloride

97%

Synonym(s):

1-(Chlorosulfonyl)-2-nitrobenzene, 2-Nitro-phenyl-sulphonyl chloride, 2-Nitrobenzene-1-sulfonyl chloride, 2-Nitrobenzenesulfonyl chloride, 2-Nitrobenzenesulphonyl chloride, 2-Nitrophenylsulfonyl chloride, o-Nitrobenzenesulfonyl chloride, o-Nitrophenylsulfonyl chloride, o-Nosyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SO2Cl
CAS Number:
Molecular Weight:
221.62
Beilstein:
521155
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

63-67 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1S(Cl)(=O)=O

InChI

1S/C6H4ClNO4S/c7-13(11,12)6-4-2-1-3-5(6)8(9)10/h1-4H

InChI key

WPHUUIODWRNJLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service