Skip to Content
Merck
All Photos(1)

Key Documents

P25507

Sigma-Aldrich

2-Phenylglycine

95%

Synonym(s):

DL-α-Phenylglycine, (±)-α-Aminophenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(NH2)COOH
CAS Number:
Molecular Weight:
151.16
Beilstein:
3197862
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white to faint yellow

mp

290 °C (subl.) (lit.)

application(s)

detection

SMILES string

NC(C(O)=O)c1ccccc1

InChI

1S/C8H9NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H,10,11)

InChI key

ZGUNAGUHMKGQNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motohiro Akazome et al.
The Journal of organic chemistry, 75(3), 660-665 (2010-01-07)
In terms of chiral recognition for racemic aryl methyl sulfoxides in the solid state, three kinds of crystalline (S)-alkylglycyl-(S)-phenylglycines were examined as potential dipeptides host molecules. When (S)-alanyl-(S)-phenylglycines [(S,S)-Ala-Phg] crystallized with aryl methyl sulfoxides, the stereochemistry of preferentially included sulfoxides
Aaron D Mills et al.
Bioorganic & medicinal chemistry letters, 20(1), 87-91 (2009-12-04)
A developing therapy of cystic fibrosis caused by the DeltaF508 mutation in CFTR employs correction of defective CFTR chloride channel gating by a 'potentiator' and of defective CFTR protein folding by a 'corrector'. Based on SAR data for phenylglycine-type potentiators
Jian-Lian Chen et al.
Analytica chimica acta, 718, 130-137 (2012-02-07)
Three different approaches for immobilizing cross-linked chitosan molecules (CS-s) in sol-gel phases to form chiral OT-CEC capillaries were comparatively investigated in this study. To synthesize column I, a bare capillary was first silanized with triethoxysilane (TEOS) and then reacted with
Shohei Tashiro et al.
Inorganic chemistry, 50(1), 4-6 (2010-12-01)
The optically active cobalt(III) complex with chiral cyclen, (2S,5S,8S,11S)-2,5,8,11-tetraethyl-1,4,7,10-tetraazacyclododecane, preferentially binds to D-phenylglycine (D-Phg) or D-t-leucine (D-t-Leu) rather than L-Phg or L-t-Leu, respectively, with 20% de in dimethyl sulfoxide at 293 K. Comparative studies on the crystal structures of cobalt(III)
James J Lynch et al.
Pain, 125(1-2), 136-142 (2006-06-20)
Gabapentin and pregabalin have been demonstrated, both in animal pain models and clinically, to be effective analgesics particularly for the treatment of neuropathic pain. The precise mechanism of action for these two drugs is unknown, but they are generally believed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service