Skip to Content
Merck
All Photos(1)

Key Documents

M68423

Sigma-Aldrich

N-Methyl-phenethylamine

99%

Synonym(s):

N-Methyl-2-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NHCH3
CAS Number:
Molecular Weight:
135.21
Beilstein:
636347
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.516 (lit.)

bp

203 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CNCCc1ccccc1

InChI

1S/C9H13N/c1-10-8-7-9-5-3-2-4-6-9/h2-6,10H,7-8H2,1H3

InChI key

SASNBVQSOZSTPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Methyl-phenethylamine can be used as a reactant:
  • To synthesize N-methyl-phenethylamine based tertiary amines by reacting with different alkyl halides in the presence of triphenylphosphine (TPP) and diisopropylazocarboxylate (DIAD) via N-alkylation reaction.
  • To fabricate photochemically stable, super-sensitive, and highly selective fluorescent film for the detection of N-methamphetamine (an illicit drug).
  • To prepare biologically active squaric acid N-hydroxylamide amide derivatives.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Efficient synthesis of tertiary amines from secondary amines
Michio K, et al.
Tetrahedron Letters, 47, 4871-4875 (2006)
Fabrication of a new fluorescent film and its superior sensing performance to N-methamphetamine in vapor phase
H Meixia, et al.
Sensors and Actuators B, Chemical, 227 (2016)
T D Forbes et al.
Journal of animal science, 72(2), 464-469 (1994-02-01)
Eighteen Suffolk and Suffolk x Hampshire wethers (56.3 +/- 1.3 kg) were used to determine the effects of naturally occurring amines, N-methyl-beta-phenethylamine (NMP) and tyramine (T), on plasma cortisol, norepinephrine (NE), ACTH, and GnRH-stimulated LH concentrations. In each experiment, wethers
B B Carpenter et al.
Journal of animal science, 72(11), 2948-2954 (1994-11-01)
The purpose of this experiment was to characterize adrenal and ovarian responses to N-methyl-beta-phenethylamine (NMP) or ACTH. Thirty-three heifers with functional midcycle corpora lutea were placed on a 4-d superovulation regimen. They were injected twice daily with FSH-P and either
Hanna-Mari Jauhonen et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 255(12), 2357-2362 (2017-08-26)
Our purpose was to investigate the effect of locally administered cis-urocanic (cis-UCA) in two experimental models of allergic conjunctivitis. The compound 48/80 (C48/80)-induced ocular irritation model (IgE-independent) and the ovalbumin (OA)-induced ocular allergy model (IgE-mediated) were used to test and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service