Skip to Content
Merck
All Photos(1)

Key Documents

M2705

Sigma-Aldrich

Mellitic acid

99%

Synonym(s):

Benzenehexacarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6(CO2H)6
CAS Number:
Molecular Weight:
342.17
Beilstein:
2228443
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

>300 °C (lit.)

SMILES string

OC(=O)c1c(C(O)=O)c(C(O)=O)c(C(O)=O)c(C(O)=O)c1C(O)=O

InChI

1S/C12H6O12/c13-7(14)1-2(8(15)16)4(10(19)20)6(12(23)24)5(11(21)22)3(1)9(17)18/h(H,13,14)(H,15,16)(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

YDSWCNNOKPMOTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Vítek et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 80(1), 32-35 (2011-02-08)
Recently, portable Raman instrumentation has been in demand for geosciences and for future planetary exploration for the identification of both organic and inorganic compounds in situ on Earth and on other planetary bodies, especially on Mars. Here we present the
J Jehlicka et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(1), 229-234 (2006-03-01)
Raman spectra have been obtained for crystals of the organic mineral mellite, from three different sites. Mellite occurs in the frame of the Tertiary series including lignite and coaly slates at Artern (Thuringia), Tula (Russia) and Bílina (Northern Bohemia). Mellite
Takeshi Nagayasu et al.
Journal of colloid and interface science, 286(2), 462-470 (2005-05-18)
Adsorption characteristics of carboxylic acids, amines, an octapeptide composed of four L-alanine and four L-aspartic acid residues (Peptide-A4D4), and beta-lactoglobulin (beta-Lg) on tantalum (Ta), titanium (Ti), and zirconium (Zr) particles were examined at 30 degrees C and in some case
Jeferson Rosário et al.
Scientific reports, 9(1), 7302-7302 (2019-05-16)
We show that filamentous fungi can emit green or red light after the accumulation of particulate lanthanide metal-organic frameworks over the cell wall. These new biohybrids present photoluminescence properties that are unaffected by the components of the cell wall. In
Alexander Apelblat et al.
The journal of physical chemistry. B, 110(17), 8893-8906 (2006-04-28)
A general approach is proposed to analyze electrical conductivities in aqueous solutions of polybasic organic acids. Experimental conductivities are examined in the context of dissociation and hydrolysis reactions by applying the Quint-Viallard conductivity equations and the Debye-Hückel equations for activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service