Skip to Content
Merck
All Photos(2)

Documents

L6375

Sigma-Aldrich

Lithium sulfate

greener alternative

≥98.5% (titration)

Synonym(s):

Dilithium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2SO4
CAS Number:
Molecular Weight:
109.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98.5% (titration)

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

845 °C (lit.)

solubility

absolute ethanol: insoluble(lit.)
acetone: insoluble(lit.)
pyridine: insoluble(lit.)

density

2.22 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li+].[Li+].[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

INHCSSUBVCNVSK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lithium sulfate is a white crystalline solid that is soluble in water. The solubility decreases with an increase in temperature. In addition, Lithium sulfate is hygroscopic in nature. It has pyroelectric properties. It has various applications in the fields of lithium-ion batteries, supercapacitors, glass industry, ceramic production, organic synthesis, and manufacture of alloys.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium sulfate can be used:
  • As a precursor to prepare solid polymer electrolytes.
  • To fabricate lithium borate glasses for energy storage applications.
  • As an electrolyte in self-healing aqueous lithium-ion batteries.
  • As an electrolyte additive in lead acid batteries. The addition of lithium sulfate improves the life cycle and efficiency of acid batteries.
  • As an electrolyte for the production of high energy and power supercapacitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Possibility of lithium sulfate borate-based glass doping with Ni and Mn ions in energy storage application
Amorntep Montreeuppathum, et al.},
Radiation Physics and Chemistry, 206, 110803-110803 (2023)
Exploring the large voltage range of carbon/carbon supercapacitors in aqueous lithium sulfate electrolyte
Qiang Gao, et al.
Energy & Environmental Science, 5, 9611-9617 (2012)
Effects of lithium sulfate and zinc sulfate additives on the cycle life and efficiency of lead acid batteries
Nnabundo N Musei, et al.
ACS Omega, 6, 4423-4429 (2021)
A self-healing aqueous lithium-ion battery
Yang Zhao, et al.},
Angewandte Chemie (International Edition in English), 55, 14384-14388 (2016)
Solid polymer electrolytes based on poly(vinylchloride)?lithium sulfate
T.Uma, et al.},
Materials Chemistry and Physics, 90, 239-244 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service