Skip to Content
Merck
All Photos(3)

Key Documents

KITALYSISBASE

Sigma-Aldrich

KitAlysis High-Throughput Base Screening Kit - Pack of 2

Synonym(s):

Base screen, Base screening kit, KitAlysis base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

functional group

phosphine

Application

Versatile base screening kit for cross-coupling reactions and other synthetic transformations. Step-by-step user guide and a downloadable experimental excel sheet are provided in the below hyperlinks.

Step-by-Step Guide for KitAlysis High-Throughput Base Reaction Screening Kit

Downloadable experimental excel file

Each kit contains 2 customizable screening sets that contain the following:
  • 7 X 2 pre-weighed bases (30 μmol) in individual microvials with magnetic stir bars
  • 4 X 2 base solutions
  • 2 empty microvials for Chemist′s Choice of base allowing customization
  • 4 degassed anhydrous solvents (DMF, MeOH, THF, Toluene)
  • 2 NEW KitAlysis 24-Well Reaction Block Replacement Films
  • 4 empty substrate stock solution reaction vials with stirbars
Monitor reaction progress via TLC-MS by following this protocol

Best when used with KitAlysis Benchtop Inertion Box and 24-well Reaction Block (sold separately).

Features and Benefits

  • Designed and tested by synthetic chemists
  • Easily run 24-microscale reactions in parallel
  • 10 μmol reactions with 3 equiv. base, 0.1 M concentration
  • Customizable Excel Template for quick calculations and easy incorporation into electronic laboratory notebooks

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules

Kit Components Also Available Separately

Product No.
Description
SDS

  • RDD019Potassium phosphate tribasic, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%SDS

  • 441902Cesium carbonate, ReagentPlus®, 99%SDS

  • 757551Potassium hydroxide, anhydrous, ≥99.95% trace metals basisSDS

  • 791733Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%SDS

  • 1390091,8-Diazabicyclo[5.4.0]undec-7-ene, 98%SDS

  • 225770Lithium bis(trimethylsilyl)amide solution, 1.0 M in THFSDS

  • 702706Sodium tert-butoxide solution, 2 M in THF, ZerO2®SDS

  • 900522Toluene, anhydrous, ZerO2®, 99.8%SDS

  • 900632Triethylamine, ZerO2®, ≥99%SDS

  • 900636Tetrahydrofuran, anhydrous, inhibitor-free, ZerO2®, ≥99.9%SDS

  • 900638N,N-Dimethylformamide, anhydrous, ZerO2®, 99.8%SDS

  • 900641Methanol, anhydrous, ZerO2®, 99.8%SDS

  • Z118850Spinbar® magnetic stirring fleas, red, L 10 mm, diam. 3 mmSDS

  • Z742110KitAlysis 24-well reaction block replacement filmSDS

  • 900501Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%SDS

  • 655104Sodium hydroxide, reagent grade, 97%, powderSDS

See All (16)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney,Adrenal gland,Testes

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

Flash Point(F)

-5.8 °F

Flash Point(C)

-21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service